CarrierMax™ FMR1 Reagent Kit
Applied Biosystems™

CarrierMax™ FMR1 Reagent Kit

The CarrierMax FMR1 Reagent Kit is used to detect the number of CGG repeats in the Fragile X Mental RetardationRead more
Have Questions?
Catalog NumberQuantity
95236248 Samples
Catalog number 952362
Price (MXN)
48,698.74
Each
Quantity:
48 Samples
Price (MXN)
48,698.74
Each

The CarrierMax FMR1 Reagent Kit is used to detect the number of CGG repeats in the Fragile X Mental Retardation (FMR1) gene. This kit uses a dual PCR system combining full length and triplet primed PCR amplification (TP-PCR), followed by fragment analysis on the Applied Biosystems 3500/3500xL Genetic Analyzer or SeqStudio Genetic Analyzer, to accurately determine up to 200 CGG repeats and detect alleles that are >200 CGG repeats.

Key features of the kit include the ability to:
• Accurately and consistently detect and classify alleles as Normal (<45), Intermediate (45-54), Premutation (55-199), and Full Mutation (≥200)
• Identify Full Mutation alleles of >200 CGG repeats
• Determine the presence of AGG interrupting sequences
• Perform fragment analysis on the Applied Biosystems 3500/3500xL Genetic Analyzer or SeqStudio Genetic Analyzer
• Analyze the results in GeneMapper Software, followed by easy reporting of allele classification using the included CarrierMax Software

Fragile X is one of the leading causes of developmental delay and one of the key targets for carrier screening in individuals planning for conception. Fragile X syndrome arises from silencing or genetic disruption of the fragile X mental retardation (FMR1) gene through increase of CGG repeats or point or structural mutations. The majority (99%) of detected aberrations are comprised of the CGG repeat expansion that increases the risk of gene silencing . The CarrierMax FMR1 Reagent Kit offers the ability to reliably and accurately screen many samples to determine the FMR1 CGG repeat status of both FMR1 copies for females and the one FMR1 gene for males. Complex samples (mosaics, XXY, etc.) can be identified for further testing.

How to use
The kit comes with enough reagents to run 48 samples for FMR1 carrier screening research. The premise behind the CGG detection is the use of a dual PCR based system using both full length and triplet repeat PCR (TP-PCR) priming methodologies.

The kit and secondary analysis software can effectively process multiple samples to full results within one working day.

Download CarrierMax Software (.zip) ›
SeqStudio GeneMapper files
3500/XL GeneMapper files

Quality of performance
The analytical sensitivity and specificity of the assay is proven to be >99% using Coriell control standards. Accuracy was determined to be >99% in sizing of CGG repeats up to 200 repeats as tested and detection of alleles with >200 repeats. Reliability of the system as determined by reproducible classifications across experiments, labs, and systems indicates the suitability of the CarrierMax FMR1 kit for carrier screening applications such as Expanded Carrier Screening research.

Specifications
Final Product Typereagent kit
For Use With (Application)Genotyping Using Fragment Analysis for Carrier Screening Research
For Use With (Equipment)Applied Biosystems 3500 Genetic Analyzer, Applied Biosystems 3500 XL Genetic Analyzer, Applied Biosystems SeqStudio Genetic Analyzer
Label or Dye6-FAM, QD1200
Product Typereagent kit
Quantity48 Samples
TargetFMR1
FormatReagent Tubes in Box
Unit SizeEach

Frequently asked questions (FAQs)

What are the recommended controls for CarrierMax FMR1 Reagent Kit (Cat. No. 952362)?

We recommend using the following controls for CarrierMax FMR1 Reagent Kit:
- NA20236: FMR1 gene (Not affected) from Coriell Institute
- NA06968: FMR1 gene (Premutation/Carrier) from Coriell Institute
- NA06897: FMR1 gene (Affected) from Coriell Institute
- NA23687: SMA1 gene (Carrier) from Coriell Institute
- NA00232: SMA1 gene (Homozygote affected) from Coriell Institute
- NA04408: SMA1 gene (Risk factor (Pathogenic SNP)) from Coriell Institute
- NA11859: (SMA1) gene (Normal) from Coriell Institute

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.

Do you have GeneMapper analysis parameters available for the CarrierMax FMR1 Reagent Kit?

We have GeneMapper analysis parameters for the CarrierMax kits, specifically for the SeqStudio and the 3500/3500xL instruments. They can be found on the following links or the CarrierMax kit product page. http://downloads.thermofisher.com/GeneMapper/FMR1_SeqStudio_GeneMapper_files.zip http://downloads.thermofisher.com/GeneMapper/FMR1_3500_GeneMapper_files.zip

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.

What software can I use to analyze data from the CarrierMax FMR1 Reagent Kit?

To analyze data from the CarrierMax FMR1 Reagent Kit, you would have to use both the GeneMapper Software and CarrierMax Software for genotyping and classification of the sample, respectively.

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.

Is the QD1200 size standard available separately from the CarrierMax FMR1 Reagent Kit?

The QD1200 size standard found in the CarrierMax FMR1 Reagent Kit is not available separately.

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.

What size standard is used in the CarrierMax FMR1 Reagent Kit?

The CarrierMax FMR1 Reagent Kit uses the QD1200 size standard.

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.