Product Image
Applied Biosystems™
Axiom™ GeneTitan™ Barcoded Stain Trays, bulk, 384HT format
Axiom GeneTitan Barcoded Stain Trays allow for processing of ten Axiom 384HT-format array plates using the GeneTitan MC instrument.
Have Questions?
Catalog NumberQuantity
9523873840 samples
Catalog number 952387
Price (MXN)
-
Quantity:
3840 samples
Axiom GeneTitan Barcoded Stain Trays allow for processing of ten Axiom 384HT-format array plates using the GeneTitan MC instrument. The barcode alerts you if an incorrect consumable has been loaded into the instrument. The barcoded stain trays need to be used with GeneTitan MC hybridization trays and scan trays/covers. The bulk packaging is ideal for high throughput processing of arrays.
Also see Axiom GeneTitan Stain Tray Covers, bulk, 384HT format
Related Links:
Specifications
No. of Samples3840 samples
Quantity3840 samples
TypeBarcoded Stain Tray
Format384 HT-array Plate
Number of Arrays3840 arrays
Unit SizeEach