Axiom™ PharmacoFocus™ Assay Core Kit, mini 96-array - FAQs
Axiom™ PharmacoFocus™ Assay Core Kit, mini 96-array - FAQs
View additional product information for Axiom™ PharmacoFocus™ Assay Core Kit, mini 96-array - FAQs (952425)
1 product FAQs found