KingFisher Apex 96 Combi tip comb (for Apex only) - FAQs
KingFisher Apex 96 Combi tip comb (for Apex only) - FAQs
View additional product information for KingFisher™ Plastics for 96 deep-well format - FAQs (97003520, 97003540, 97003530, 97002570, 97002534B, 97002534, 97003500)
4 product FAQs found
Do you offer an alternative for the discontinued Pharma MagMAX Express-96 Deep-Well Tip Combs (Cat. No. A31537)?
Yes, we recommend using KingFisher 96 tip combs for deep-well format (Cat. No. 97002534 or 97002570).
Are KingFisher Plastics for 96-deep well format autoclavable?
No, KingFisher Plastics for 96-deep well format are not autoclavable. They won't melt, but they will get deformed.
Can I reuse the Kingfisher Plastics for 96 deep-well format?
The Kingfisher Plastics for 96 deep-well format are recommended for one-time use only, and we don't recommend reusing these products.
Are the KingFisher Plastics for 96 deep-well format CE marked?
The KingFisher Plastics for 96 deep-well format are not CE marked.