KingFisher 96 Deep-well Tip Comb and Plate, sterile, Barcoded (for Apex) - FAQs
KingFisher 96 Deep-well Tip Comb and Plate, sterile, Barcoded (for Apex) - FAQs
View additional product information for KingFisher 96 Deep-well Plates - FAQs (95040450, 95040460, 95040460B, 97002820B, 97002820, 95040450B, 95040450BU)
1 product FAQs found
I received the KingFisher Flex/KingFisher Duo Prime tip combs and the retaining clips are bent or broken off. When I try to unbend the retaining clips, they break. Can I still use the tip combs if the retaining clips are bent or broken off?
Do not use the tip combs if the clips are bent or missing. Not having clips can cause the tip comb to drop from the holder during the run, which would cause the run to abort. If you received tip combs with bent or broken retaining clips, you can contact Technical Support at techsupport@thermofisher.com to request a free-of-charge replacement.
Find additional tips, troubleshooting help, and resources within our Automated Nucleic Acid Purification Support Center.