When should I use a GeneArt Gene Synthesis Kit, order GeneArt Strings DNA Fragments, or use GeneArt Gene Synthesis services?
The kit and custom services offer different advantages. Please click here (http://www.thermofisher.com/us/en/home/life-science/cloning/gene-synthesis/geneart-gene-synthesis-product-selection-guide.html) to view a selection guide comparing the technologies, including deliverables and production time, to determine what is best for you.
What is gene optimization and how does it differ from codon optimization?
Our proprietary GeneOptimizer software (https://www.thermofisher.com/us/en/home/life-science/cloning/gene-synthesis/geneart-gene-synthesis/geneoptimizer.html) calculates the optimal DNA sequence needed to encode the protein of interest (gene optimization). Adapting the codon usage of the gene to the codon preferences of the expression organism (codon optimization) is just one of the parameters addressed. The GeneOptimizer software currently evaluates several additional parameters that can compromise mRNA stability, such as extreme GC content, ribosomal binding sites, consensus and cryptic splice sites, repeats, and secondary structures. Increased numbers of stable mRNA molecules often lead to higher yields of protein.
What does your GeneArt gene synthesis service include?
GeneArt Gene Synthesis Service includes gene optimization (if requested), synthesis of the DNA, cloning into our standard vector, and delivery of 5 µg of lyophilized DNA plus a compact disc containing the in silico analysis and quality control information.
Can I have my gene delivered in my vector?
Yes. We can subclone your gene into a vector you provide. You will receive 5 µg each of your gene in a GeneArt cloning vector and in your vector (plus the quality control information for both constructs).
Can I use pMx for my expression studies?
Unfortunately, this vector does not contain expression elements such as a promoter or terminator for proper protein expression. We recommend subcloning your synthetic gene into an expression vector of your choice via restriction enzyme cloning, or we can perform this for you as a custom service.
Do you offer a kit for site-directed mutagenesis?
Yes, we offer three kits for site-directed mutagenesis: GeneArt Site-Directed Mutagenesis System and GeneArt Site-Directed Mutagenesis PLUS kit, Phusion Site-Directed Mutagenesis Kit as well as a custom GeneArt service.
The kits are almost identical, but the PLUS kit contains an improved 2x GeneArt Enzyme mix with protocols optimized for multi-site mutagenesis (up to 3 sites in plasmids up to 14 kb). The PLUS kit can also be used for single-site mutations of up to 25 nucleotides, and the original kit can be used for single substitution/deletion/insertions up to 12 nucleotides in plasmids up to 14 kb. A web tool was also introduced for primer design, but this can also be used for the original kit. Phusion Site-Directed Mutagenesis Kit is a versatile and efficient tool for introducing point mutations, insertions, or deletions in any type of plasmid DNA. With this kit, the entire plasmid is amplified using phosphorylated primers that introduce the desired changes. The amplified, linear PCR product, containing the desired mutation, is circularized in a 5-minute ligation reaction with T4 DNA Ligase. The resulting plasmid can be then transformed into any competent E. coli cells.
I'd like to use the GeneOptimizer process to optimize my gene, but need to protect a specified area of my sequence. Can this be done?
Yes, you can protect specified regions or restriction enzymes sites that you would not like to be optimized by the the GeneOptimizer process. In the portal, under "Optimize sequence", choose "Protect Restriction Sites" and/or "Protect Sequence Parts" from optimization.
What ori is in the GeneArt pMx standard vectors?
ColE1 is the ori.
Can I choose the restriction enzyme sites used for cloning into your standard pMx vector?
Due to our production process, this is not an option we can offer to you. Restriction enzymes are added that generally eliminate most of the standard vectors' multiple cloning site.
What vector will my gene get cloned into when using GeneArt Gene Synthesis Services?
Genes are typically cloned into one of our GeneArt pMx standard vectors. These vectors contain
• a multiple cloning site
• an Ori (colE1)
• an antibiotic resistance marker (ampicillin, kanamycin, streptomycin, chloramphenicol); please note standard orders do not guarantee any specific standard vector
What makes a gene complex?
There are multiple factors to be considered: CG content, repetition score, direct
repeats, and secondary structures, as well the assigned host organism and total length of the gene. All of these things are taken into consideration when deciding whether or not a gene or a part of a sequence is complex'.
I want to order my gene with the Superspeed option, but it is greyed out. Why?
The Superspeed delivery option is only available for non-complex genes, up to 3 kb in size. If your gene does not fit these requirements, the option will be greyed out.
What is a variant and what are the guidelines for variants? What are the criteria or requirements for single variants and/or variant bundles of a master/parent gene?
Single variants and variant bundles (more than one variant process) are variants of a master gene. The variant criteria are as follows:
Variants may contain up to four of the following modifications:
- 5'/3' deletion of any length with additional 52 nt of new sequence on each side
- 5'/3' modification of a maximum of 52 nt on each side
- 5'/3' extension of a maximum of 52 nt on each side
- Internal modifications of up to 40 nt (a maximum of 3 of these internal modification blocks are allowed)
- Internal deletions of any length
- Modifications (whether internal or 5´/3´) must be separated by at least 100 bp.
If a gene falls outside of these criteria, it must be created as a separate gene synthesis instead of as a variant.
Can I get my synthetized gene subcloned into a TOPO vector?
Currently, we do not offer this as a service. Subcloning can be achieved using a restriction enzyme–based vector or via BP/LR clonase reactions into a Gateway vector. Alternatively, the GeneArt Strings DNA Fragments we offer can be directly cloned into a TOPO Zero Blunt vector.
What do I receive when I order a gene synthesis project through GeneArt services?
You will receive 5µg lyophilized plasmid (synthesized gene cloned into pMx plasmid), unless otherwise requested. Upon reciept, the DNA can be re-suspended and stored in aliquots at –20 degrees C. You will also receive a CD with sequencing alignment data along with your DNA.
What is the size of my pMX vector?
The sizes vary slightly due to the size of the anitibiotic resistance marker. pMA:2.5kb, pMS: 2.6kb, pMC: 2.3kb, pMK: 2.4kb.
What factors does the GeneOptimizer process optimize for my synthetic gene?
The GeneOptimizer process takes a multi-parameter approach to analyze the gene sequence. It will take codon usage, GC content, cryptic splice sites, direct repeats, RNA secondary structures, and instability sequences into account when optimizing a gene sequence.
Is there a size limit for my gene synthesis?
In theory, no there is no limit. However, very large DNA fragments (greater than 10 kb) need to be cloned into a low-copy vector to ensure genetic stability. Therefore, increasing length leads to increased production time.
I do not want my GeneArt synthetic gene to be cloned into a vector. Can I just get it delivered as a double stranded fragment?
We offer a GeneArt Strings DNA Fragments service, which provides custom-made linear, double-stranded DNA fragments up to 1,000 bp in length. Go to www.thermofisher.com, and search for "GeneArt Strings DNA Fragments" to learn more about this product.
Are there standard primer sites that I can use in your pMx vector for sequencing?
Yes, all pMX vectors contain an M13-20 primer binding site and a M13-R primer binding site. The sequences are as follows:
M13-20 forward primer binding site: 5' - TTGTAAAACGACGGCCAG - 3'br/>
M13-R primer binding site: 5' - GGAAACAGCTATGACCATGT - 3'
What is the difference between the GeneArt standard vectors, i.e., pMA, pMC, pMK, pMS, pMZ?
pMx is the standard GeneArt cloning vector used to clone in your synthetic gene of interest. The x stands for the antibiotic of choice. Therefore, pMA stands for ampicillin, pMK stands for kanamycin, pMS stands for spectinomycin, pMC stands for chloramphenicol, and pMZ stands for Zeocin antibiotic. All vectors contain a multiple cloning site, an ORI (colE1), and an antibiotic resistance marker, along with M13-20/M13-R primer binding sites for sequencing.