Search
Search

GeneArt Custom Gene Synthesis Services |
GeneArt custom gene synthesis service offers a reliable and cost-effective method for tailoring DNA sequences with 100% accuracy. In addition, we offer sequence optimization with our patented GeneOptimizer algorithm which uses a multifactorial approach that goes beyond basic codon optimization to maximize gene expression.
Custom gene constructs can be ordered online using the GeneArt Services Dashboard or via email. The GeneArt Services Dashboard is our proprietary platform that allows you to optimize protein-coding sequences, design constructs, generate quotes, manage orders, and import templates.
Features of GeneArt Services Dashboard:
For more details and tutorials, visit the GeneArt Dashboard online guide.
Order through the Classic GeneArt Gene Synthesis Portal
Download the sequence submission form for email ordering
Download New GeneArt Dashboard User Manual in Korean
Typical production times are shown for complex and non-complex sequences in the table below. Sequence complexity is determined automatically when designing constructs online and takes into consideration several factors including GC-content, sequence repeats, homopolymers, and secondary structure.
Please note: For the best pricing and shorter production time, optimize your sequences to reduce complexity.
| Gene length (bp) | Production time (business days) | |
|---|---|---|
| Non-complex sequence | Complex sequence | |
| 100–1,000 | 5–8 | 5–10 |
| 1,001–3,000 | 5–8 | 12+ |
| 3,001–5,000 | 7–12 | 12+ |
| 5,001–7,000 | 12+ | 12+ |
| 7,001–12,000 | N/A | As quoted |
Options to get your cloned genes even faster
Synthetic genes are provided as clones in the customer vector of choice to allow for flexibility depending on downstream application. The final plasmid is shipped vacuum dried in 2 mL tubes and ready to resuspend in water or an appropriate buffer. The standard delivery amount is 5 µg, however, scaled-up plasmid preparations are available as an add-on service if ready-to-use plasmid DNA is desired.
De novo genes synthesized by GeneArt Services can be cloned into a standard shuttle vector, pMX, or a variety of popular vectors.
The vector onboarding option is useful when the gene synthesis vector selection tool does not include your desired vector, or if you need a synthesized gene delivered in your own vector.
De novo genes synthesized by GeneArt Services can be cloned into a standard shuttle vector, pMX, or a variety of popular vectors.
The vector onboarding option is useful when the gene synthesis vector selection tool does not include your desired vector, or if you need a synthesized gene delivered in your own vector.
Watch how-to videos and explainers on GeneArt Instant Designer.
Contact us
Request more information
North America and Asia: 1-800-955-6288
Ask for "GeneArt" when prompted
Europe: +49 (0)941 942 76-100
Order support:
Geneartsupport@thermofisher.com
Stay up to date on gene synthesis and protein synthesis service offerings.
Read white papers on de novo gene synthesis technologies.
![]()
For Research Use Only. Not for use in diagnostic procedures.