Gallery Reagents for Food and Beverage Analysis
Gallery Reagents for Food and Beverage Analysis
Actual product may vary
Gallery Reagents for Food and Beverage Analysis
Gallery Reagents for Food and Beverage Analysis
Thermo Scientific™

Gallery Reagents for Food and Beverage Analysis

Choose from a continuously expanding selection of tests for automated food and beverage analysis.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeQuantity
984304D to Glucose System Reagent20 mL Vials
984300Ethanol System Reagent20 mL Vials
984302D to Fructose System Reagent20 mL Vials
981842Wash Fluid20 mL Vials
984332ECM pH 7 Standard Test20 mL Vials
984353BC System Liquid20 mL Vials
984354BC Diluent20 mL Vials
984355BC Eluent20 mL Vials
984764D to Glucose Reagent20 mL Vials
984765Glycerol20 mL Vials
984305Beta to Glucan System Reagent2 Reagents in 20 mL Vials
984306D to Lactic Acid System Reagent20 mL Vials
984307Potassium System Reagent1 Reagent in 20 mL Vial
984308L to Lactic Acid System Reagent20 mL Vials
984309Tartaric Acid System Reagent20 mL Vials
984310L to Malic Acid System Reagent20 mL Vials
984312Sucrose System Reagent20 mL Vials
984314D to Glucose + D to Fructose System Reagent20 mL Vials
984315Lipase System Reagent20 mL Vials
984316Glycerol System Reagent20 mL Vials
984317D to Glucose + D to Fructose + Sucrose System Reagent20 mL Vials
984318Acetic Acid System Reagent20 mL Vials
984319L to Asparagine/L to Aspartic Acid20 mL Vials
984320Ammonia System Reagent20 mL Vials
984321Urea System Reagent20 mL Vials
984322D to Isocitric Acid System Reagent20 mL Vials
984323Lactose System Reagent20 mL Vials
984324Cholesterol in Food System Reagent20 mL Vials
984326Total Iron System Reagent2 Reagents in 20 mL Vials, 1 Reagent Capsule
984327Citric Acid System Reagent2 Working Reagents in 20 mL Vials
984328Total Protein System Reagent1 Reagent in 20 mL Vial
984342Alpha to Amino Nitrogen2 Reagents in 20 mL Vials
984345Total Sulfite System Reagent2 Reagents in 20 mL Vials
984346Total Polyphenol System Reagent2 Reagents in 20 mL Vials
984347Acetaldehyde System Reagent2 Reagents in 20 mL Vials
984348Oxalic Acid System Reagent2 Reagents in 20 mL Vials
984349pH (Colorimetric) System Reagent1 Reagent in 20 mL Vial
984356Acetic Acid System Reagent2 Working Reagents in 20 mL Vials
984380Sugar Combination Standard Reagent3 x 3 mL
984382Acid Combination Standard Reagent3 x 3 mL
984383Beta to Glucan Standard Reagent1 x 50 mg
984384Ethanol Standard Reagent10 x 1 mL
984386Glycerol Standard Reagent3 x 3 mL
984388Urea Standard Reagent3 x 3 mL
984390Lactose Standard Reagent3 x 3 mL
984391Cholesterol in Food Standard Reagent3 x 3 mL
984393Oxalic Acid Standard Reagent3 x 3 mL
984394NOPA Standard Reagent3 x 3 mL
984396Acetaldehyde Standard3 x 5 mL
984632Total Acids (Wine pH 7)1 buffer and 1 color reagent
984633Total Acids (Juice pH 8)1 buffer and 1 color reagent
984634Free Sulfite System Reagent2 Reagents in 20 mL Vials
984635L to Ascorbic Acid System Reagent3 Working Reagents in 20 mL Vials
984636L-glutamic Acid System Reagent20 mL Vials
984710D to Gluconic Acid System Reagent2 Working Reagents in 20 mL Vials
984373Alpha Amylase System ReagentReagent A: 6 x 20 mL Vials
Reagent B: 2 x 5 mL Vials
Catalog number 984304
Price (USD)
-
Request A Quote
Type:
D to Glucose System Reagent
Quantity:
20 mL Vials
Save time by eliminating reagent preparation with the Thermo Scientific™ Gallery™ system reagents for food and beverage analysis. Over 50 ready-to-use gallery system reagents are offered for food and beverage analysis, with the kits including two to four reagents in 20 mL barcoded vials. This enables the necessary identification data, such as lot numbers and expiration dates, to be read automatically by the discrete analyzer's barcode reader. Automated dilutions make it possible to apply methods for a wide range of analyte concentrations. Save time and reduce errors by simplifying your analysis with the Gallery system reagents.
  • Kit structure includes 2 to 4 reagents in 20mL vials
  • Barcoded vials are simple and fast to use
  • All necessary data, such as lot and expiration date, is automatically read
  • Photometric measurement is mainly based on the formation or consumption of NAD or NADH
  • Measurements are done at 340nm in 37°C
  • Automated dilutions make it possible to apply methods for a wide range of analyte concentrations
Specifications
Analytical MethodEnzymatic test with hexokinase and glucose-6-phosphate dehydrogenase
DescriptionD-Glucose system reagents (300 tests)
DetectsD-Glucose
Interference Filter340 / 600
No. of Tests300
Quantity20 mL Vials
Test Range0.1 to 160 g/L, 5 to 500 mg/L
Sample TypeFood and Beverage Sample
TypeD to Glucose System Reagent
Unit SizeEach