Gallery™ Plus Discrete Analyzer
Actual product may vary
Thermo Scientific™
Gallery™ Plus Discrete Analyzer
Save time and cost with a fully automated, high-capacity, benchtop photometric analyzer specifically developed for food, beverage, water and soil testing.
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호설명
98620001Gallery Plus Discrete Analyzer without ECM Unit
98621001Gallery Plus Discrete Analyzer with ECM Unit
카탈로그 번호 98620001
제품 가격(KRW)
-
설명:
Gallery Plus Discrete Analyzer without ECM Unit
Experience simultaneous analysis of multiple analytes with the discrete cell technology of the Thermo Scientific™ Gallery™ Plus discrete analyzer. This fully automated, high-capacity analyzer provides fast, reproducible results and has an integrated platform that offers two measurement techniques, photometric (colorimetric and enzymatic) and electrochemical (pH and conductivity) analysis, that can be operated in parallel for analytical efficiency. The Gallery Plus automated discrete analyzer can accommodate 108 samples and 42 reagents in its separated sample and reagent disks, with the capability to perform as many as 350 tests per hour. Achieve superior analytical performance with a wide range of applications including food and beverage analysis, environmental testing, and industrial quality control.
Automated operation and reliable technology
  • Continuous access to samples, reagents, and cuvettes without interruption of routine workflow.
  • Integrated platform for photometric and electrochemical analysis can be operated simultaneously.
  • Increased throughput with a sample capacity of up to 350 tests/hour.
  • All necessary analysis steps are automated to reduce operator error.
  • Up to 2 hours walk-away time, significantly reducing operator time.
  • Dilutions and repeat analyses are handled automatically.
  • Minimal sample pretreatment required.
Robust capability
  • Combined sample and reagent disk with up to 180 sample positions and reagent disk for 42 reagents.
  • Measurements cover a large concentration range with excellent reproducibility.
  • User-specific applications can be developed to meet individual testing needs.
  • Several blanking possibilities to eliminate sample matrix effects.
  • Ability to achieve detection of very low levels (ppb).
  • Wavelength range from 340nm to 880nm.
  • Sophisticated dilution management.
Compact design and economical
  • System reagents eliminate time-consuming reagent preparation, freeing operators for other tasks.
  • Unique low volume cuvette design for less reagent waste and potentially lowering operation costs.
  • Contamination-free analysis using disposable cuvettes.
  • Optional ECM unit for conductivity and pH analysis.
  • No need for external water source.
Versatile and user-friendly
  • Accommodates a variety of sample cups and primary tubes.
  • No risk of carry-over or requirement to wash glassware.
  • No additional priming or method changeover time.
  • Automated identification by barcode reader.
  • Self-contained benchtop for small footprint.
  • Automatic startup and shutdown protocols.
  • Intuitive graphical user-interface.
사양
보정Factor, Bias, Linear, Logit-Log, Spline, Polynomial and Point-to-Point
큐벳Discrete Disposable Cuvettes. Capacity 360 Measurement Cells.
깊이(영국식 단위)27.5 in.
깊이(미터법)70 cm
설명Gallery Plus Discrete Analyzer without ECM Unit
증류수 소비Deionized Water Consumption 2.5 L/hr.
주파수50/60 Hz +/-5%
높이(영국식 단위)24.4/51.1 in. (Cover Closed/Cover Open)
높이(미터법)62/130 cm (Cover Closed/Cover Open)
측정 방법Photometric
운영 체제Automated Photometric (Colorimetric and Enzymatic) Analysis
품질 관리 세부 정보Real-Time QC Program with Multiple and Variable Rules. Control Frequency User-Definable. Out-of-Specification Control Results Flagged.
시약9- or 18-Position Sample Racks, Max 108 Positions (= 6 Racks) on-Board; Reagent Capacity 42 Positions on-Board
샘플 희석Automatic Sample Pre-Dilution. Automatic Sample Dilution of Over-Range Tests with Automatic re-Run. Addition of Manual Pre-Dilution Value for Result Calculation.
처리량Up to 350 Tests/hr.
전압100/240 V +/-10%
중량(미터법)110 kg
폭(영국식 단위)37 in.
폭(미터법)94 cm
샘플 종류Food and Beverage Sample, Environmental Sample
유형Photometric Analyzer
Unit SizeEach
자주 묻는 질문(FAQ)
Can I integrate discrete analysis with ion chromatography for comprehensive water analysis?
Yes. We offer Disc IC packages that pair discrete analysis with Thermo Scientific Dionex ion chromatography systems, delivering high throughput, comprehensive water/ion analysis without method changeover.
How does a discrete analyzer compare to flow analyzers (FIA/SFA/CFA)?
Compared with traditional continuous-flow systems, Gallery and Gallery Plus discrete analyzers’ random-access, discrete design with disposable cuvettes minimizes carryover, cuts reagent use (typically <100 μL per test), shortens startup/changeover times, and boosts throughput (200–350 tests/hr vs 60–120 tests/hr). Labs also report much lower cost-per-test (up to 20x) due to smaller volumes and less waste. These systems also provide more measuring channels (12 filter positions), greater method stability, and fully automated micro liquid handling, which reduce operator skill requirements and system maintenance.
What reagents and consumables are available? Can I use third party reagents?
We offer ready to use, barcoded system reagents and enzymatic kits for environmental/industrial water and food & beverage testing, these minimize prep time, consumption, and cost per analysis. The platform is also open and flexible to support third party photometric methods and reagents if needed.
What “walk away” and automation features are built in?
After loading samples and reagents, the system can run unattended for up to 3 hours. Automation features include the ability to add samples mid-cycle (for the next cycle) without disrupting current tests, automated start up/shut down, and barcode scanning/LIMS connectivity for traceability.
Can I run photometric tests and pH/conductivity at the same time?
Yes. With the optional Electrochemical Measurement (ECM) module, pH (approximately 2 to 12) and conductivity (~20 μS/cm–112 mS/cm) can be measured in parallel with standard photometric assays, enabling truly consolidated multiparameter runs. ECM throughput is up to 54 measurements/hr on Gallery systems and up to 67 measurements/hr on Gallery Plus models.