Gallery™ Plus Aqua Master Discrete Analyzer
Gallery™ Plus Aqua Master Discrete Analyzer
Gallery™ Plus Aqua Master Discrete Analyzer
Thermo Scientific™

Gallery™ Plus Aqua Master Discrete Analyzer

Maximize productivity with the cost-effective, fully automated Thermo Scientific™ Gallery™ Plus Aqua Master for reliable and reproducible water, nutrient and agricultural testing.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberElectrochemistry Unit (ECM)
98620005Optional
98621005Included
Catalog number 98620005
Price (USD)
-
Electrochemistry Unit (ECM):
Optional

Maximize your productivity with the Thermo Scientific™ Gallery™ Plus Aqua Master discrete analyzer, a high-throughput, fully automated, discrete photometric analyzer for water, nutrient and agricultural testing. It generates fast, reproducible results in a compact, benchtop design and can run different analyses simultaneously from a single sample to help reduce total analysis time and cost. Combining rugged hardware and custom-designed software, Gallery Aqua Master helps you meet regulatory requirements while automating complicated calibration sequences, quality control, and spiking procedures.

Automated operation and reliable technology
  • Advanced software features including choice of calibration order, relative standard error calculation, automated spiking procedures, automated QC procedures, flexible reporting, and smart suggested dilutions
  • Automated water and nutrient analysis workflows that meet U.S. EPA guidelines and other local regulations
  • Continuous access to samples, reagents, and cuvettes without interruption of routine workflows
  • Integrated platform for simultaneous photometric and electrochemical analysis
  • Automates all required analysis steps, enabling up to 3 hours of walkaway time and reducing manual errors
  • Increased throughput with a sample capacity of up to 350 tests/hour
Robust capabilities
  • Separate sample and reagent disks with up to 108 sample positions on board and 42 reagent positions
  • Detection levels in the ppb range with excellent reproducibility
  • Twelve filter positions and wavelength range from 340 nm to 880 nm
  • Long-life xenon light source good for years of operation
  • Disposable cuvettes minimize potential for contamination
  • Low maintenance with less than one service visit required per year, on average
Versatile and economical
  • Ready-to-use system reagents eliminate time-consuming reagent preparation
  • Uniquely low-volume cuvettes for lowest reagent consumption and waste and up to 20 times lower operating costs
  • Open system allows you to modify an existing method or create your own test, and adopt 3rd party reagents
User-friendly operation
  • Suitable for all user expertise levels with minimal training required
  • Automatic startup and shutdown protocols
  • Self-contained benchtop for small footprint
Specifications
Analytical MethodDiscrete Analysis
DescriptionGallery™ Plus Aqua Master without ECM Unit
Dimensions (D x W x H)70 x 94 x 62/130 cm (cover closed/cover open)
Electrochemistry Unit (ECM)Optional
For Use With (Application)Water Analysis, Nutrient Analysis, Environmental Testing, Industrial Water Testing, Agricultural Testing
Incubation Temperature25°C to 60°C
Measurement MethodPhotometric: 340 nm–880 nm
pH Range: 3–7.2
Reagent CapacityUp to 42 reagent positions on board
Sample SizeUp to 108 sample positions on board
ThroughputUp to 350 Photometric Tests/h
Weight (Metric)110 kg
TypeDiscrete Analyzer
Unit SizeEach

Frequently asked questions (FAQs)

Does the Gallery Aqua Master discrete analyzer support US EPA / NELAC methods, and what about nitrate/nitrite (TON) without cadmium?

Yes. Gallery Aqua Master software automates methods that comply with US EPA, NELAC, and other international standards. For TON, the platform supports EPA approved enzymatic reduction methods, a safer alternative to cadmium coil reductions, while automating the required calibrations and QC to meet method criteria.

Which analytes can the Gallery Aqua Master discrete analyzer measure, and how many at once?

The platform can run up to 20 parameters simultaneously from a single sample. Common parameters include ammonia, nitrate + nitrite (TON), nitrite, orthophosphate, silica, chloride, sulfate, total phosphorus (TP), total Kjeldahl nitrogen (TKN), hexavalent chromium, cyanide, total hardness, metals (colorimetric), and pH & conductivity using the optional Electrochemical Measurement (ECM) module.

How is the Gallery Aqua Master different from the standard Thermo Scientific Gallery discrete analyzer?

Both systems use our discrete cell technology, but the Gallery Aqua Master system adds water focused software capabilities: pre defined environmental/water QC method panels and custom software features for regulated methods (automated spiking, flexible QC, regulatory style calibrations, smart dilutions, and reporting). The standard Gallery discrete analyzer is a versatile general purpose discrete analyzer for food, beverage, environmental, and industrial applications, whereas the Gallery Aqua Master system is purpose built for water and nutrient compliance workflows.

What is the Gallery Aqua Master system?

The Thermo Scientific Gallery Aqua Master system is a fully automated, discrete photometric analyzer designed specifically for water and nutrient testing. It automates wet chemistry methods (colorimetric/enzymatic) to deliver reproducible results for environmental, industrial and agricultural labs that comply with US EPA, NELAC, and other international regulatory standards while reducing manual steps and errors.

Can I integrate discrete analysis with ion chromatography for comprehensive water analysis?

Yes. We offer Disc IC packages that pair discrete analysis with Thermo Scientific Dionex ion chromatography systems, delivering high throughput, comprehensive water/ion analysis without method changeover.