Indiko™ Plus Clinical Chemistry Analyzer
Thermo Scientific™

Indiko™ Plus Clinical Chemistry Analyzer

Experience an uncompromised level of performance and ease of use with the Thermo Scientific™ Indiko™ Plus Clinical Chemistry Analyzer.
Have Questions?
Catalog NumberDescription
98640000Indiko Plus without ISE Unit
Catalog number 98640000
Price (USD)
-
Description:
Indiko Plus without ISE Unit

Experience an uncompromised level of performance and ease of use with the Thermo Scientific™ Indiko™ Plus Clinical Chemistry Analyzer. This fully-automated random access benchtop system for clinical and specialty chemistry testing, combined with system reagents and consumables form a fully-supported, complete system solution offering confidence to daily work.

  • IVDR-compliant, cost-effective solution for small and medium laboratory settings
  • Methods used in Indiko Plus are traceable to international reference methods and materials
  • Sample oriented testing generates results quickly, enhancing the quality of patient care
  • Application protocols include routine clinical chemistry tests as well as special tests, such as specific proteins, drug of abuse testing, therapeutic drug monitoring and immunosuppressant drug testing
  • Simple and quick daily start-up and maintenance procedures
  • Continuous access to samples, cuvettes and reagents without interrupting the testing process
  • Disposable cuvettes allow user-safe and contamination free analysis
  • Versatile, automated dilution features make sample processing easier
  • Reflex testing capability offers savings in operator time, triggering automatically the specific test to be run
  • Measurement reliability can be monitored with a real-time QC program
  • Automatic upload of calibrator and control values save time and reduce manual errors
  • Results archiving associated with calibration and reagent lot data secure full traceability
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  

Specifications

  • Operating Principle: Fully automated, sample oriented, random access analyzer
  • Capacity: photometric tests/hr: up to 350 ; ISE tests/hr: up to 135
  • On-board capacity: up to 108 sample positions and 42 reagent positions
  • Sample volume: 2 to 120μL
  • Reagent volume: 2 to 240μL (1 to 4 reagent additions/test)
  • Reaction volume: 120 to 300μL
  • Incubator: 9 x 10-position single-use cuvettes, totally 90 cuvette positions
  • Photometer: 12 filter positions
  • Absorbance range: 0 to 3.5A
  • Water consumption: 2.5L/hr.
  • Workstation: separate PC
  • Bi-directional LIS-connection
  • Power consumption: 300w
  • Dimensions (L x W x H): 27.6 x 37 x 24.4in. (70 x 94 x 62cm)
  • Weight: 242 lb. (110kg)
Specifications
Analytical MethodClinical and Specialty Chemistry Testing with Random Access Analysis
CapacityCapacity: photometric tests/hr: up to 350; On-board capacity: up to 108 sample positions and 42 reagent positions
CuvettesDisposable Cuvette
DescriptionIndiko Plus without ISE Unit
Distilled Water Consumption2.5 L/hr.
Height (English)24.4 in.
Height (Metric)62 cm
QuantityEach
ReagentsReagent volume: 2 to 240µL (1 to 4 reagent additions/test)
Weight (English)242 lb.
Weight (Metric)110 kg
Width (English)37 in.
Width (Metric)94 cm
TypeClinical Chemistry Analyzer
Unit SizeEach