Cytidine, 99%
Cytidine, 99%
Cytidine, 99%
Thermo Scientific Chemicals

Cytidine, 99%

Used to synthesize enzyme inhibitors, antiviral agents, and anticancer agents. And also used as constituent of nucleic acids. | CAS: 65-46-3 | C9H13N3O5 | 243.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number A10261.09
Price (USD)
75.70
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
75.70
Each
Chemical Identifiers
CAS65-46-3
IUPAC Name4-amino-1-[3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2-dihydropyrimidin-2-one
Molecular FormulaC9H13N3O5
InChI KeyUHDGCWIWMRVCDJ-UHFFFAOYNA-N
SMILESNC1=NC(=O)N(C=C1)C1OC(CO)C(O)C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
Assay (Non-aqueous acid-base Titration)≥98.5 to ≤101.5% (non-U.S. specification)
Assay from Suppliers CofA≥98.5% (HPLC: U.S. specification)
CommentSpecification differs for U.S. and non-U.S. material where indicated
View more
Cytidine is used to synthesize enzyme inhibitors, antiviral agents, and anticancer agents. And also used as constituent of nucleic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only