FoamAway™ Irradiated AOF (animal origin-free) Antifoaming Agent, 2.5L in a 5L Bag - FAQs
FoamAway™ Irradiated AOF (animal origin-free) Antifoaming Agent, 2.5L in a 5L Bag - FAQs
View additional product information for FoamAway™ Irradiated AOF (animal origin-free) Antifoaming Agent - FAQs (A1036902, A1036901)
4 product FAQs found
What is the tolerance for the target simethicone concentration in FoamAway Irradiated AOF (animal origin-free) Antifoaming Agent and what is the maximum concentration it can have?
The target shipping percentage of simethicone in FoamAway Irradiated AOF (animal origin-free) Antifoaming Agent is 3%. This supports the defoaming assay noted on the COA for the shelf life of the product.
FoamAway Irradiated AOF (animal origin-free) Antifoaming Agent is formulated to a 4.0% simethicone concentration during manufacture, representing the upper end of the product's concentration range, to account for loss of simethicone during processing.
The final concentration of simethicone may be variable in the finished product, but is ≤4.0%. The final product must pass a defoaming activity assay as a quality release criterion, ensuring defoaming activity throughout the 8-month shelf life.
Find additional tips, troubleshooting help, and resources within our Transfection Support Center.
Why does the FoamAway Irradiated AOF Antifoaming Agent (Cat. No. A1036901, A1036902) have a shelf life of 12 months? Do you have any data on extending the expiry time frame?
Thermo Fisher Scientific conducted extended stability testing showing that FoamAway Irradiated AOF Antifoaming Agent remains within specification for 12 months from the date of irradiation. At the 13-month time point, multiple lots met the defoaming activity specification (≤ 15 seconds). Thus, 12 months is the supported shelf life based on validated stability data./
What type of connectors are used in FoamAway Irradiated AOF (animal origin-free) Antifoaming Agent?
The connectors are male and female luer locks.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What is your usage recommendation for FoamAway Irradiated AOF (animal origin-free) Antifoaming Agent?
We recommend shaking to thoroughly mix the product before each addition. Approximately 1:1,000 dilution may be the midrange of the working concentration, about 30 ppm. The range may be about a 1:20,000 to 1:600 depending on the process. For an extended culture, we recommend re-adding material during the run as it is incorporated with the foaming components; we recommend adding as little as possible to keep the downstream processes functioning.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.