AbC™ Anti-Rat/Hamster Bead Kit
Invitrogen™

AbC™ Anti-Rat/Hamster Bead Kit

The AbC™ Anti-Rat/Hamster Antibody Compensation Bead Kit provides a consistent, accurate, and simple-to-use technique for the setting of flow cytometryRead more
Have Questions?
Catalog NumberQuantity
A103891 kit
Catalog number A10389
Price (USD)
429.00
Each
Quantity:
1 kit
Price (USD)
429.00
Each
The AbC™ Anti-Rat/Hamster Antibody Compensation Bead Kit provides a consistent, accurate, and simple-to-use technique for the setting of flow cytometry compensation when using fluorochrome-conjugated to rat or hamster antibodies with any laser in flow cytometry.

• Species reactivity—for use with all isotypes of rat and hamster antibody conjugates
• Maximum laser compatibility—for use with all lasers

Accurate compensation is crucial for effective multicolor analysis in flow cytometry. Often, however, cellular-based compensation controls are not completely effective, as many antigens are not highly expressed, and dimly stained cells can lead to inaccurate compensation settings. The AbC™ Anti-Rat/Hamster Bead Kit was developed to address this issue.

View a selection guide for all flow cytometry compensation beads.

How it works
Each kit provides polystyrene microspheres that have fluorescent antibody conjugate capture capacity (positive compensation beads) or are inert (negative compensation beads). After mixing with a fluorophore-conjugated rat or hamster antibody, the two components provide a distinct high-signal positive control with an appropriate negative population that can then be used to set compensation properly regardless of the intensity of the cells in the actual experiment.

Species reactivity
The AbC™ Anti-Rat/Hamster Bead Kit is ideal for compensation with all isotypes of rat (IgG1, IgG2a, IgG2b, IgG2c, IgM) and hamster (Armenian IgG and Syrian IgG) immunoglobulins.

Maximum laser compatibility
All AbC™ Compensation Bead Kits are ideal for use with any excitation source, including the 405 nm laser which has historically provided higher background signals due to auto-fluorescence with other kits on the market.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodFluorescence
Diameter (Metric)6 μm
Emissionany lasers (UV to 633/635)
For Use With (Equipment)Flow Cytometer
FormatTube
Quantity1 kit
Shipping ConditionRoom Temperature
Stain ConfigurationSurface
Product LineCompenFlow, Molecular Probes
Product TypeCompensation Beads
Unit SizeEach
Contents & Storage
Contains 1 vial AbC™ anti-rat/hamster capture beads and 1 vial of negative beads.
  • Store at 2°C to 8°C
  • Frequently asked questions (FAQs)

    Does the AbC Anti-Rat/Hamster Bead Kit bind IgG2a kappa or IgG1 lambda rat with the same affinity?

    Sorry, we have not analyzed this.

    Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

    I am performing flow cytometry. My compensation matrix has values over 100. That's not possible is it?

    It is possible. But if you have a lot of values over 100, you probably need to look at the voltages that you are using for your data collection.

    Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

    I’ve been told that I need to compensate my data for flow cytometry analysis. What does that mean?

    By running single-color controls, it is possible to remove signal of one fluorophore that spills over into the collection channel for another fluorophore.

    Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

    What kind of controls do I need for flow cytometry?

    For compensation, you need to prepare a singly stained sample (or compensation beads) for each color parameter that you are using. In addition, we recommend that you use FMO (flow minus one) controls. These are controls in which you label cells or beads with every color in your panel, omitting one. Make one FMO control for each color. These controls are important for helping you properly set gates on your data.

    Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

    Why should I worry about compensation in flow cytometry analysis?

    In a perfect world, the fluorescence emission profile for each individual fluorophore would be a very intense, narrow peak, well separated from all other emission peaks. In reality, organic dyes and fluorescent proteins have broad emission peaks, and compensation must be employed (during or after data acquisition) to correctly assign fluorescence signal to each fluorophore. Compensation is important because it removes fluorescent signal from overlapping spectra so you know that the signal you see is only the signal from the fluorophore of interest.

    Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

    Citations & References (3)

    Citations & References
    Abstract
    Identification and characterization of intestinal antigen-presenting cells involved in uptake and processing of a nontoxic recombinant chimeric mucosal immunogen based on cholera toxin using imaging flow cytometry.
    Authors:Zhao W, Minderman H, Russell MW,
    Journal:
    PubMed ID:24197893
    'Intragastric immunization with recombinant chimeric immunogen, SBR-CTA2/B, constructed from the saliva-binding region (SBR) of Streptococcus mutans antigen AgI/II and the A2/B subunits of cholera toxin (CT) induces salivary and circulating antibodies against S. mutans that protect against dental caries. We previously found that SBR-CTA2/B activated dendritic cells (DC) in the ... More
    Multiparametric analysis of host response to murine cytomegalovirus in MHC class I-disparate mice reveals primacy of Dk-licensed Ly49G2+ NK cells in viral control.
    Authors:Prince J, Lundgren A, Stadnisky MD, Nash WT, Beeber A, Turner SD, Brown MG,
    Journal:
    PubMed ID:24068668
    MHC class I D(k) and Ly49G2 (G2) inhibitory receptor-expressing NK cells are essential to murine CMV (MCMV) resistance in MA/My mice. Without D(k), G2(+) NK cells in C57L mice fail to protect against MCMV infection. As a cognate ligand of G2, D(k) licenses G2(+) NK cells for effector activity. These ... More
    Self MHC class I-licensed NK cells enhance adaptive CD8 T-cell viral immunity.
    Authors:Stadnisky MD, Xie X, Coats ER, Bullock TN, Brown MG
    Journal:Blood
    PubMed ID:21436069
    MHC class I (MHC I) is essential to NK- and T-cell effector and surveillance functions. However, it is unknown whether MHC I polymorphism influences adaptive immunity through NK cells. Previously, we found that MHC I D(k), a cognate ligand for the Ly49G2 inhibitory receptor, was essential to NK control of ... More