L-Histidine, 98+%
L-Histidine, 98+%
L-Histidine, 98+%
Thermo Scientific Chemicals

L-Histidine, 98+%

CAS: 71-00-1 | C6H9N3O2 | 155.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A10413.14
also known as A10413-14
Price (USD)
32.65
Special offer
Online exclusive
Ends: 30-Jun-2026
38.80
Save 6.15 (16%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
32.65
Special offer
Online exclusive
Ends: 30-Jun-2026
38.80
Save 6.15 (16%)
Each
Chemical Identifiers
CAS71-00-1
IUPAC Name2-amino-3-(1H-imidazol-5-yl)propanoic acid
Molecular FormulaC6H9N3O2
InChI KeyHNDVDQJCIGZPNO-UHFFFAOYNA-N
SMILESNC(CC1=CN=CN1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (Non-aqueous acid-base Titration)≥98.0 to ≤102.0%
Optical Rotation-39 ? 2? (c = 5 in water)
Identification (FTIR)Conforms
Appearance (Color)White
L-Histidine acts as a precursor to histamine and a component of carnosine. It is also used in medicine, feed additive, biochemical research, dietary supplement. It is used in a nutrition enhancer, is the amino acid fluid infusion. It can be used in the treatment of gastric ulcer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Histidine acts as a precursor to histamine and a component of carnosine. It is also used in medicine, feed additive, biochemical research, dietary supplement. It is used in a nutrition enhancer, is the amino acid fluid infusion. It can be used in the treatment of gastric ulcer.

Solubility
Soluble in water (15 mg/ml at 20°C).

Notes
Store at 4°C.Incompatible with oxidizing agents.
RUO – Research Use Only
JJ Madden; EL McGandy; NC Seeman. The crystal structure of the orthorhombic form of L-(+)-histidine. Acta Cryst. 1972, B28, 2377-2382.