Alexa Fluor™ 594 Hydrazide, for microinjection - FAQs
Alexa Fluor™ 594 Hydrazide, for microinjection - FAQs
View additional product information for Alexa Fluor™ 594 Hydrazide, for microinjection - FAQs (A10442)
1 product FAQs found
What dye can I use that is non-reactive and can show an injection site?
The non-reactive Alexa Fluor and Alexa Fluor hydrazide derivatives may be used for injection site visualization. Other options include the fluorescent polystyrene microspheres, FluoSpheres, and dye-conjugated dextrans. The hydrazide derivatives and 'fixable' dextrans are retained by cross-linking using an aldehyde-based fixative.