CTS™ OpTmizer™ T Cell Expansion SFM, bag format
Gibco™
CTS™ OpTmizer™ T Cell Expansion SFM, bag format
Gibco™ OpTmizer™ CTS™ T-Cell Expansion SFM has been developed for the growth and expansion of human T lymphocytes. OpTmizer™ CTS™자세히 알아보기
Have Questions?
카탈로그 번호수량
A10485031 Bag
카탈로그 번호 A1048503
제품 가격(KRW)
491,000
Each
수량:
1 Bag
제품 가격(KRW)
491,000
Each
Gibco™ OpTmizer™ CTS™ T-Cell Expansion SFM has been developed for the growth and expansion of human T lymphocytes. OpTmizer™ CTS™ T-Cell Expansion medium is a complete serum-free and xeno-free 1X medium consisting of OpTmizer™ T-Cell Expansion Basal Medium (1 L bag) and OpTmizer™ T-Cell Expansion Supplement (26 mL), which are mixed together prior to use.
  • Maintains similar phenotype and function as T-cells cultured in conventional serum supplemented medium
  • Consistent performance in supporting T-cell expansion
  • Supports high density cultures in static and WAVE Bioreactor™ cultures
  • Be ready for clinical research with this cGmP manufactured formula and support documentation
Versatile T Cell Culture Media
OpTmizer™ CTS™ T-Cell Expansion SFM is designed to give results equivalent to conventional serum-supplemented media. However, if desired, 2% heat-inactivated human serum (Cat. No. 34005-100) may be added to the medium to enhance viability and expansion. The use of serum, and the amount required, should be determined empirically depending on the specific T-cell culture application.
Ready for T Cell Expansion
OpTmizer™ CTS™ T-Cell Expansion SFM is designed for the expansion of human T-cells permitting CD3+ densities of >3x106 cells/mL in static conditions and >2x107 cells/mL in WAVE bags. Results may vary depending on the T-cell subset (e.g., Ag-specific cells). Conditions for specific T-cell culture applications should be determined empirically.
Stability of Complete Media
Complete OpTmizer™ CTS™ T-Cell Expansion SFM (basal, supplement, and L-glutamine) is stable for 4 weeks when stored in the dark at 2–8°C.
Simplify Your Transition into Clinical Research
The Gibco™ Cell Therapy Systems (CTS™) brand offers a broad array of products for use in cell therapy applications, including media, reagents, growth factors, enzymes, cell selection/expansion reagents, and devices. Gibco™ CTS™ products deliver:
  • Reduced burden in qualifying reagents
  • Appropriate certificates of analyses and origin included
  • Convenient access to our Drug Master File (DMF)
사양
세포 유형T Cells
배양 환경CO2
엔도톡신 수준Very Low
항생제 포함No antibiotics
무기 염류Calcium, Magnesium
제품 유형Cell Expansion Serum Free Medium (SFM)
순도 또는 품질 등급Research Grade
수량1 Bag
분류Chemically-defined, Serum-free, Xeno-free
Culture TypeSuspension Cell Culture
형태Liquid
Serum LevelSerum-free
멸균Sterile
첨가제 포함HEPES, Phenol Red, Sodium Pyruvate, Sodium Bicarbonate
첨가제 없음No Glutamine
Unit SizeEach
구성 및 보관
OpTmizer™ T-Cell Expansion Basal Medium (1 x L media bag): Store at 2–8°C. Protect from light.
OpTmizer™ T-Cell Expansion Supplement (1 x 26 mL): Store in the dark at 2–8°C.
자주 묻는 질문(FAQ)
What is CTS?
The Gibco Cell Therapy Systems (CTS) portfolio of cell and gene therapy products are GMP manufactured, safety tested, and backed by regulatory documentation to support your transition from discovery through clinical and commercial manufacturing. Through our CTS solutions, we are committed to helping customers streamline therapeutic development, minimize risk, and ease the burden on their quality systems.
Do I need to order additional components with the CTS OpTmizer T Cell Expansion SFM (Cat. Nos. A1048501, A1048502, A1048503)?
The CTS OpTmizer T Cell Expansion SFM (Cat. Nos. A1048501, A1048502, A1048503) kit comes with OpTmizer T Cell Expansion Basal Medium and OpTmizer T Cell Expansion Supplement, which are mixed together prior to use. Depending on your protocol you may need to purchase T cell growth factors such as IL-2 (Cat. No. PHC0023) and L-glutamine (Cat. No. 25030) to form a complete medium. To boost growth of human T cells, human AB serum can be added. If desired, antibiotics such as Gentamicin (Cat. No. 15750), can also be added.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Will CTS OpTmizer T Cell Expansion SFM (Cat. No. A1048501, A1048502, A1048503) work with all types of T cells?
CTS OpTmizer T Cell Expansion SFM (Cat. No. A1048501, A1048502, A1048503) has been demonstrated to be effective for the ex vivo expansion of human CD3 T cells, including polyclonal T cell populations, antigen-specific T cells, tumor-infiltrating lymphocytes (TIL), cord blood T cells, regulatory T cells, T cells from HIV-infected donors, gene-modified T cells (both lenti- and MLV-based vectors), including cells of both the CD4+ and CD8+ phenotype.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Can CTS OpTmizer T Cell Expansion SFM (Cat. Nos. A1048501, A1048502, A1048503) be used for animal T cell cultures other than human?
CTS OpTmizer T Cell Expansion SFM (Cat. No. A1048501, A1048502, A1048503) has only been tested on human T cells.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Is there a difference in formulation of the CTS OpTmizer T Cell Expansion SFM between the bottle and bag packaging formats?
The bottle and bag packaging formats of the medium have the same formulation.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
인용 및 참조 문헌 (5)
인용 및 참조 문헌
Abstract
Ex vivo expansion of human T cells for adoptive immunotherapy using the novel Xeno-free CTS Immune Cell Serum Replacement.
Authors:Smith C, Økern G, Rehan S, Beagley L, Lee SK, Aarvak T, Schjetne KW, Khanna R
Journal:
PubMed ID:25671129
The manufacture of clinical grade cellular products for adoptive immunotherapy requires ex vivo culture and expansion of human T cells. One of the key components in manufacturing of T cell therapies is human serum (HS) or fetal bovine serum (FBS), which can potentially expose immunotherapy recipient to adventitious infectious pathogens ... More
Bioengineering and serum free expansion of blood-derived ?d T cells.
Authors:Sutton KS, Dasgupta A, McCarty D, Doering CB, Spencer HT
Journal:Cytotherapy
PubMed ID:27260209
'Cellular immunotherapy relies on several highly variable patient-specific parameters, such as (i) cell number before and after expansion, (ii) targeting of cells to tumors, (iii) cell survival and function after infusion, and (iv) on- and off-target adverse events. Cellular approaches such as the specific expansion of ?d T cells as ... More
A Rapid Cell Expansion Process for Production of Engineered Autologous CAR-T Cell Therapies.
Authors:Lu TL, Pugach O, Somerville R, Rosenberg SA, Kochendefer JN, Better M, Feldman SA
Journal:Hum Gene Ther Methods
PubMed ID:27897048
The treatment of B-cell malignancies by adoptive cell transfer (ACT) of anti-CD19 chimeric antigen receptor T cells (CD19 CAR-T) has proven to be a highly successful therapeutic modality in several clinical trials.(1-6) The anti-CD19 CAR-T cell production method used to support initial trials relied on numerous manual, open process steps, ... More
Enhancing adoptive cancer immunotherapy with V?2Vd2 T cells through pulse zoledronate stimulation.
Authors:Nada MH, Wang H, Workalemahu G, Tanaka Y, Morita CT
Journal:J Immunother Cancer
PubMed ID:28239463
Human ?d T cells expressing V?2Vd2 T cell receptors monitor foreign- and self-prenyl pyrophosphate metabolites in isoprenoid biosynthesis to mediate immunity to microbes and tumors. Adoptive immunotherapy with V?2Vd2 T cells has been used to treat cancer patients with partial and complete remissions. Most clinical trials and preclinical studies have ... More
Specific humoral and cellular immune responses in cancer patients undergoing chronic immunization with a VEGF-based therapeutic vaccine.
Authors:Morera Y, Sánchez J, Bequet-Romero M, Selman-Housein KH, de la Torre A, Hernández-Bernal F, Martín Y, Garabito A, Piñero J, Bermúdez C, de la Torre J, Ayala M, Gavilondo JV
Journal:Vaccine
PubMed ID:28536029
CIGB-247 is a cancer therapeutic vaccine, based on recombinant modified human vascular endothelial growth factor (VEGF) as antigen, in combination with the adjuvant VSSP, a bacterially-derived adjuvant. The vaccine have demonstrated efficacy in several murine malignancy models. These studies supported the rationale for a phase I clinical trial where safety, ... More