Search
View additional product information for StemPro™ MSC SFM XenoFree - FAQs (A1067501)
1 product FAQs found
StemPro MSC SFM XenoFree contains 2.2 g/L of sodium bicarbonate.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.