Tali™ Apoptosis Kit - Annexin V Alexa Fluor™ 488 & Propidium Iodide - FAQs
Tali™ Apoptosis Kit - Annexin V Alexa Fluor™ 488 & Propidium Iodide - FAQs
View additional product information for Tali™ Apoptosis Kit - Annexin V Alexa Fluor™ 488 & Propidium Iodide - FAQs (A10788)
1 product FAQs found
What are the fluorescence excitation/emission maxima for the dyes contained in the Tali Apoptosis Kit - Annexin V Alexa Fluor 488 & Propidium Iodide?
Annexin V Alexa Fluor 488: 488/499 nm
Propidium Iodide: 535/617 nm, bound to DNA