alpha-D-Lactose monohydrate
alpha-D-Lactose monohydrate
alpha-D-Lactose monohydrate
Thermo Scientific Chemicals

alpha-D-Lactose monohydrate

CAS: 5989-81-1 | C12H24O12 | 360.312 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
5000 g
Catalog number A11074.0I
also known as A11074-0I
Price (USD)
238.65
Special offer
Online exclusive
Ends: 30-Jun-2026
281.00
Save 42.35 (15%)
Each
Quantity:
5000 g
Request bulk or custom format
Price (USD)
238.65
Special offer
Online exclusive
Ends: 30-Jun-2026
281.00
Save 42.35 (15%)
Each
Chemical Identifiers
CAS5989-81-1
IUPAC Name(2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-{[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol hydrate
Molecular FormulaC12H24O12
InChI KeyWSVLPVUVIUVCRA-KPKNDVKVSA-N
SMILESO.OC[C@H]1O[C@@H](O[C@H]2[C@H](O)[C@@H](O)[C@@H](O)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Water Content (Karl Fischer Titration)4.5-5.5% (0.9-1.1 waters, theory 5.0%)
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
Optical Rotation+53.5° ±2.5° (c=10 in water/NH₃)
alpha-D-Lactose monohydrate is used as a carrier and stabiliser of aromas, pharmaceutical products, Food industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water.

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Gavin M. Walker.; Clive R. Holland.; , Mohammad M.N. Ahmad.; Duncan Q.M. Craig. Influence of process parameters on fluidised hot-melt granulation and tablet pressing of pharmaceutical powders. Chem. Eng. Sci.. 2005, 60 (14),3867-3877.
  2. Kamlesh C. Patel.; Xiao Dong Chen. Drying of aqueous lactose solutions in a single stream dryer. Food and Bioproducts Processing. 2008, 86(3), 185-197.