Bac-to-Bac™ N-His TOPO™ Expression System - FAQs
Bac-to-Bac™ N-His TOPO™ Expression System - FAQs
View additional product information for Bac-to-Bac™ N-His TOPO™ Expression System - FAQs (A11101)
1 product FAQs found
Which strand is produced using M13K07 to make ssDNA from pPROEX HT or pFastBAC vectors?
You will get the antisense strand with pProEXHT.
You will get the + or sense strand with pFastBac I of pFastBac HT.