Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • beta Galactosidase Antibodies

          Invitrogen

          beta Galactosidase Polyclonal Antibody

          46 Published Figures
          121 References
          View all (64) beta Galactosidase antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite beta Galactosidase Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Published Figures (46)
          beta Galactosidase Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          beta Galactosidase Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 46

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          beta Galactosidase Antibody (A-11132) in ICC/IF

          Cells 2021 - Figure 4 Confocal microscopy images show an increased expression of beta-galactosidase (green fluorescence) as the relative fluorescence units/area at 4 weeks but not at 12 weeks ( Table 2 ). Podocalyxin (red fluorescence) identifies the podocytes within the glomerulus. Unilateral nephrectomy is associated with glomerular hypertrophy. The bar graph shows the net relative fluorescence units/glomerulus. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          beta Galactosidase Antibody in Immunocytochemistry (ICC/IF)
          beta Galactosidase Antibody in Immunocytochemistry (ICC/IF)
          beta Galactosidase Antibody in Immunocytochemistry (ICC/IF)
          beta Galactosidase Antibody in Immunocytochemistry (ICC/IF)
          beta Galactosidase Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          beta Galactosidase Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          beta Galactosidase Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          beta Galactosidase Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          beta Galactosidase Antibody in Immunocytochemistry (ICC/IF)
          beta Galactosidase Antibody in Immunohistochemistry, Immunohistochemistry (Paraffin) (IHC, IHC (P))
          beta Galactosidase Antibody in Immunohistochemistry, Immunohistochemistry (Paraffin) (IHC, IHC (P))
          beta Galactosidase Antibody in Immunohistochemistry, Immunohistochemistry (Paraffin) (IHC, IHC (P))
          beta Galactosidase Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          beta Galactosidase Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          beta Galactosidase Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          beta Galactosidase Antibody in Immunohistochemistry (Frozen) (IHC (F))
          beta Galactosidase Antibody in Immunohistochemistry (Frozen) (IHC (F))
          beta Galactosidase Antibody in Immunohistochemistry (IHC)
          beta Galactosidase Antibody in Immunohistochemistry (IHC)
          beta Galactosidase Antibody in Immunohistochemistry (IHC)
          beta Galactosidase Antibody in Immunohistochemistry (IHC)
          beta Galactosidase Antibody in Immunohistochemistry (IHC)
          beta Galactosidase Antibody in Immunohistochemistry (IHC)
          beta Galactosidase Antibody in Immunohistochemistry (IHC)
          beta Galactosidase Antibody in Immunohistochemistry (IHC)
          beta Galactosidase Antibody in Immunohistochemistry (IHC)
          beta Galactosidase Antibody in Immunohistochemistry (IHC)
          beta Galactosidase Antibody in Immunohistochemistry (IHC)
          beta Galactosidase Antibody in Immunohistochemistry (IHC)
          beta Galactosidase Antibody in Immunohistochemistry (IHC)
          beta Galactosidase Antibody in Immunohistochemistry (IHC)
          beta Galactosidase Antibody in Immunohistochemistry (IHC)
          beta Galactosidase Antibody in Immunohistochemistry (IHC)
          beta Galactosidase Antibody in Immunohistochemistry (IHC)
          beta Galactosidase Antibody in Immunohistochemistry (IHC)
          beta Galactosidase Antibody in Immunohistochemistry (IHC)
          beta Galactosidase Antibody in Immunohistochemistry (IHC)
          beta Galactosidase Antibody in Immunohistochemistry (IHC)
          beta Galactosidase Antibody in Western Blot (WB)
          beta Galactosidase Antibody in Western Blot (WB)
          beta Galactosidase Antibody in Western Blot (WB)
          beta Galactosidase Antibody in Western Blot (WB)
          beta Galactosidase Antibody in Western Blot (WB)
          beta Galactosidase Antibody in Western Blot (WB)
          beta Galactosidase Antibody in Western Blot (WB)
          beta Galactosidase Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          A-11132

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          Assay-dependent
          View 14 publications 14 publications

          Immunohistochemistry (IHC)

          Assay-dependent
          View 74 publications 74 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          -
          View 8 publications 8 publications

          Immunohistochemistry (Frozen) (IHC (F))

          -
          View 6 publications 6 publications

          Immunocytochemistry (ICC/IF)

          Assay-dependent
          View 14 publications 14 publications

          ELISA (ELISA)

          Assay-dependent
          View 1 publication 1 publication

          ChIP assay (ChIP)

          -
          View 2 publications 2 publications

          Miscellaneous PubMed (Misc)

          -
          View 2 publications 2 publications
          Product Specifications

          Species Reactivity

          Bacteria

          Published species

          Bacteria, Fruit fly, Hamster, Human, Mouse, Rat, Tag

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          beta-Galactosidase

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          2 mg/mL

          Amount

          1 mg

          Purification

          IgG fraction

          Storage buffer

          PBS, pH 7.2

          Contains

          5mM sodium azide

          Storage conditions

          4°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_221539

          Product Specific Information

          The E. coli gene lacZ encoding beta-galactosidase is an important reporter gene for detecting the expression of recombinant genes in animal cells. In situations where enzymatic detection of beta-galactosidase is not practical, anti-beta-galactosidase antibody has been widely used. Anti-beta-galactosidase antibody can also be used for purification of fusion proteins by immunoprecipitation or immunoadsorption. Furthermore, anti-beta-galactosidase antibody has been shown to be the preferred analytical tool for light microscopic evaluation of lacZ gene transfer, and the beta-galactosidase/anti-beta-galactosidase complex has been used as a model to assess the feasibility of a high-pressure immunodesorption process.

          This antibody, raised in rabbit against E. coli beta-galactosidase, demonstrates high specificity for the enzyme and is suitable for immunological methods that require high titer and specificity such as immunoblotting (western or dot blot), ELISA, immunoprecipitation, and immunohistochemistry. The appropriate dilution of antibody for a particular application should be determined empirically. We suggest trying dilutions of about 1:5,000 to 1:50,000 for your initial experiments.

          Target Information

          Beta Galactosidase encodes a member of the glycosyl hydrolase 35 family of proteins. Alternative splicing results in multiple transcript variants, at least one of which encodes a preproprotein that is proteolytically processed to generate the mature lysosomal enzyme. This enzyme catalyzes the hydrolysis of a terminal beta-linked galactose residue from ganglioside substrates and other glycoconjugates. Mutations in this gene have been associated with GM1-gangliosidosis and Morquio B syndrome. Diseases associated with GLB1 include Gm1-Gangliosidosis, Type I and Gm1-Gangliosidosis, Type Ii.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: Beta-Gal; betagal; bgal; β-gal; β-galactosidase; βgal

          View more View less

          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-svw7g:80/100.66.72.150:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch