Can the Hibernate-E Medium (Cat. No. A1247601) be used for plating and growing mouse primary hippocampal neurons?
Hibernate-E medium (Cat. No. A1247601) is designed for the maintenance of embryonic neural tissue and is suitable for short-term maintenance of cultured rat neurons at 37°C in ambient CO2 (up to 2 days) and long-term storage of viable brain tissue at 2 to 8°C (up to a month).
However, it is not recommended for the initial plating and growing of neurons.