Albumin from Bovine Serum (BSA), Alexa Fluor™ 488 conjugate
Invitrogen™

Albumin from Bovine Serum (BSA), Alexa Fluor™ 488 conjugate

This albumin from bovine serum (BSA), Alexa Fluor 488 conjugate is a bright and photostable conjugate exceptionally suited for applicationsRead more
Have Questions?
Catalog NumberQuantity
A131005 mg
Catalog number A13100
Price (USD)
296.02
Special offer
Online exclusive
Ends: 31-Dec-2026
361.00
Save 64.98
Each
Quantity:
5 mg
Price (USD)
296.02
Special offer
Online exclusive
Ends: 31-Dec-2026
361.00
Save 64.98
Each
This albumin from bovine serum (BSA), Alexa Fluor 488 conjugate is a bright and photostable conjugate exceptionally suited for applications in which well-defined molecular weight tracers are needed. These applications include studying the rate of endocytosis and exocytosis and following intracellular protein processing.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Label TypeAlexa Fluor Dyes
Product LineAlexa Fluor
Protein SubtypeAlbumin Proteins
Quantity5 mg
Shipping ConditionRoom Temperature
ConjugateAlexa Fluor 488
FormLyophilized
SpeciesBovine
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -30°C) and protect from light.

Citations & References (25)

Citations & References
Abstract
Differential requirement for classic and novel PKC isoforms in respiratory burst and phagocytosis in RAW 264.7 cells.
Authors:Larsen EC, DiGennaro JA, Saito N, Mehta S, Loegering DJ, Mazurkiewicz JE, Lennartz MR
Journal:J Immunol
PubMed ID:10946313
'The binding of Ab (IgG)-opsonized particles by FcgammaRs on macrophages results in phagocytosis of the particles and generation of a respiratory burst. Both IgG-stimulated phagocytosis and respiratory burst involve activation of protein kinase C (PKC). However, the specific PKC isoforms required for these responses have yet to be identified. We ... More
Gbetagamma activation of Src induces caveolae-mediated endocytosis in endothelial cells.
Authors:Shajahan AN, Tiruppathi C, Smrcka AV, Malik AB, Minshall RD,
Journal:J Biol Chem
PubMed ID:15345719
'Caveolae-mediated endocytosis in endothelial cells is stimulated by the binding of albumin to gp60, a specific albumin-binding protein localized in caveolae. The activation of gp60 induces its cell surface clustering and association with caveolin-1, the caveolar-scaffolding protein. This interaction leads to G(i)-induced Src kinase activation, which in turn signals dynamin-2-mediated ... More
Regulation of caveolar endocytosis by syntaxin 6-dependent delivery of membrane components to the cell surface.
Authors:Choudhury A, Marks DL, Proctor KM, Gould GW, Pagano RE
Journal:Nat Cell Biol
PubMed ID:16565709
'Caveolar endocytosis has an important function in the cellular uptake of some bacterial toxins, viruses and circulating proteins. However, the molecular machinery involved in regulating caveolar uptake is poorly defined. Here, we demonstrate that caveolar endocytosis is regulated by syntaxin 6, a target membrane soluble N-ethylmaleimide attachment protein receptor (t-SNARE) ... More
Concentration and separation of proteins in microfluidic channels on the basis of transverse IEF.
Authors:Macounová K, Cabrera CR, Yager P
Journal:Anal Chem
PubMed ID:11321320
'The use of microfluidic channels formed by two electrodes made of gold or palladium to perform transverse isoelectric focusing (IEF) is presented as a means for continuous concentration and fractionation of proteins. The microchannels were 40 mm long with an electrode gap of 1.27 mm and a depth of 0.354 ... More
Carbon nanotubes as intracellular protein transporters: generality and biological functionality.
Authors:Kam NW, Dai H
Journal:J Am Chem Soc
PubMed ID:15839702
'Various proteins adsorb spontaneously on the sidewalls of acid-oxidized single-walled carbon nanotubes. This simple nonspecific binding scheme can be used to afford noncovalent protein-nanotube conjugates. The proteins are found to be readily transported inside various mammalian cells with nanotubes acting as the transporter via the endocytosis pathway. Once released from ... More