Albumin from Bovine Serum (BSA), Alexa Fluor™ 594 conjugate
Bovine serum albumin, Alexa Fluor 594 conjugate is a bright and photostable conjugate exceptionally suited for applications in which well-definedWeitere Informationen
Have Questions?
Katalognummer
Menge
A13101
5 mg
Katalognummer A13101
Preis (EUR)
480,00
Each
Menge:
5 mg
Preis (EUR)
480,00
Each
Bovine serum albumin, Alexa Fluor 594 conjugate is a bright and photostable conjugate exceptionally suited for applications in which well-defined molecular weight tracers are needed. These applications include studying the rate of endocytosis and exocytosis and following intracellular protein processing.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
MarkertypAlexa Fluor Dyes
ProduktlinieAlexa Fluor
Protein-SubtypAlbumin Proteins
Menge5 mg
VersandbedingungRoom Temperature
KonjugatAlexa Fluor 594
FormLyophilized
SpeziesBovine
Unit SizeEach
Inhalt und Lagerung
Store in freezer (-5 to -30°C) and protect from light.
Zitierungen und Referenzen (16)
Zitierungen und Referenzen
Abstract
Gbetagamma activation of Src induces caveolae-mediated endocytosis in endothelial cells.
Authors:Shajahan AN, Tiruppathi C, Smrcka AV, Malik AB, Minshall RD,
Journal:J Biol Chem
PubMed ID:15345719
'Caveolae-mediated endocytosis in endothelial cells is stimulated by the binding of albumin to gp60, a specific albumin-binding protein localized in caveolae. The activation of gp60 induces its cell surface clustering and association with caveolin-1, the caveolar-scaffolding protein. This interaction leads to G(i)-induced Src kinase activation, which in turn signals dynamin-2-mediated ... More
Albumin mediates the transcytosis of myeloperoxidase by means of caveolae in endothelial cells.
Authors:Tiruppathi C, Naqvi T, Wu Y, Vogel SM, Minshall RD, Malik AB,
Journal:Proc Natl Acad Sci U S A
PubMed ID:15136724
'Myeloperoxidase (MPO), the phagocyte hemoprotein involved in neutrophil host defense and consuming nitric oxide (*NO), induces the nitration of extracellular matrix proteins and tissue remodeling subsequent to its transcytosis across the endothelial barrier. We addressed the role of an interaction of MPO with albumin as a requirement for MPO transport ... More
Concentration and separation of proteins in microfluidic channels on the basis of transverse IEF.
Authors:Macounová K, Cabrera CR, Yager P
Journal:Anal Chem
PubMed ID:11321320
'The use of microfluidic channels formed by two electrodes made of gold or palladium to perform transverse isoelectric focusing (IEF) is presented as a means for continuous concentration and fractionation of proteins. The microchannels were 40 mm long with an electrode gap of 1.27 mm and a depth of 0.354 ... More
Real-time differential labeling of blood, interstitium, and lymphatic and single-field analysis of vasculature dynamics in vivo.
'Lymph nodes are highly organized structures specialized for efficient regulation of adaptive immunity. The blood and lymphatic systems within a lymph node play essential roles by providing functionally distinct environments for lymphocyte entry and egress, respectively. Direct imaging and measurement of vascular microenvironments by intravital multiphoton microscopy provide anatomical and ... More
Initiation and termination of NF-kappaB signaling by the intracellular protozoan parasite Toxoplasma gondii.
Authors:Shapira S, Harb OS, Margarit J, Matrajt M, Han J, Hoffmann A, Freedman B, May MJ, Roos DS, Hunter CA,
Journal:J Cell Sci
PubMed ID:16079291
Signaling via the NF-kappaB cascade is critical for innate recognition of microbial products and immunity to infection. As a consequence, this pathway represents a strong selective pressure on infectious agents and many parasitic, bacterial and viral pathogens have evolved ways to subvert NF-kappaB signaling to promote their survival. Although the ... More