L-Cystine, 99%
L-Cystine, 99%
L-Cystine, 99%
Thermo Scientific Chemicals

L-Cystine, 99%

CAS: 56-89-3 | C6H12N2O4S2 | 240.292 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1000 g
Catalog number A13762.0B
also known as A13762-0B
Price (USD)
376.65
Special offer
Online exclusive
Ends: 30-Jun-2026
443.00
Save 66.35 (15%)
Each
Quantity:
1000 g
Request bulk or custom format
Price (USD)
376.65
Special offer
Online exclusive
Ends: 30-Jun-2026
443.00
Save 66.35 (15%)
Each
Chemical Identifiers
CAS56-89-3
IUPAC Name(2R)-2-amino-3-{[(2R)-2-amino-2-carboxyethyl]disulfanyl}propanoic acid
Molecular FormulaC6H12N2O4S2
InChI KeyLEVWYRKDKASIDU-IMJSIDKUSA-N
SMILESN[C@@H](CSSC[C@H](N)C(O)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
Assay (Non-aqueous acid-base Titration)≥98.5 to ≤101.5%
Identification (FTIR)Conforms
Optical Rotation-205° to -224° (c=1 in 1N HCl)
L-Cystine is used as an antioxidant, protecting tissues against radiation and pollution. It finds application in protein synthesis. It is required for utilization of vitamin B6 and is useful in healing burns and wounds. It is also is required by certain malignant cell lines in the culture medium as well as for the growth of certain micro-organisms. It is useful in the stimulation of hematopoietic system and promotes the formation of white and red blood cells. It is an active ingredient in medications used to treat dermatitis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Chen, L.; Zheng, Z.; Wang, J.; Wang, X. Mesoporous SBA-15 end-capped by PEG via L-cystine based linker for redox responsive controlled release. Microporous Mesoporous Mater. 2014, 185, 7-15.
  2. Liu, M.; Zhang, L.; He, X.; Zhang, B., Song, H.; Li, S.; You, W. l-Cystine-assisted hydrothermal synthesis of Mn 1- x Cd x S solid solutions with hexagonal wurtzite structure for efficient photocatalytic hydrogen evolution under visible light irradiation. J. Mater. Chem. A 2014, 2 (13), 4619-4626.