Geltrex™ Reduced-Growth Factor Basement-Membrane Matrix, LDEV-free, stem-cell qualified, 5 mL - FAQs
Geltrex™ Reduced-Growth Factor Basement-Membrane Matrix, LDEV-free, stem-cell qualified, 5 mL - FAQs
View additional product information for Geltrex™ Reduced-Growth Factor Basement-Membrane Matrix, LDEV-free, stem-cell qualified - FAQs (A1413302)
16 product FAQs found
Can I use the Geltrex hESC-Qualified, Ready-To-Use, Reduced Growth Factor Basement Membrane Matrix (Cat. No. A1569601) with other PSC media systems (i.e. StemFlex, Essential 8, Essential 8 Flex, etc.)?
No, the Ready-To-Use Geltrex matrix was developed for use with the StemPro hESC SFM System. It is not recommended for other PSC media systems.
For other PSC media systems, we recommend the Geltrex LDEV-Free, hESC-Qualified, Reduced Growth Factor Basement Membrane Matrix (Cat. Nos. A1413301, A1413302).
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What is the difference between Geltrex LDEV-Free and Geltrex hESC-Qualified, Ready-To-Use, Reduced Growth Factor Basement?
Geltrex hESC-Qualified, Ready-To-Use, Reduced Growth Factor Basement Membrane Matrix (Cat. No. A1569601) is a pre-diluted Geltrex product ready to use for coating culture vessels. There is no need to dilute the solution further. The product has also been QC tested using hESCs to ensure that every lot will maintain the attached pluripotency of these cells. The full product description can be found here:
Geltrex hESC-Qualified, Ready-To-Use, Reduced Growth Factor Basement Membrane Matrix (Cat. No. A1569601)
Geltrex LDEV-Free, hESC-Qualified, Reduced Growth Factor Basement Membrane Matrix (Cat. Nos. A1413301, A1413302) is a "concentrated" version of the Geltrex hESC-Qualified, Ready-To-Use above. You can pre-dilute this concentrated solution to pre-coat the culture vessel to grow hESCs at your desired usage concentration. This Geltrex matrix has also been QC tested using hESCs to ensure every lot will maintain the attached pluripotency of these cells. The full product description can be found here:
Geltrex LDEV-Free, hESC-Qualified, Reduced Growth Factor Basement Membrane Matrix (Cat. Nos. A1413301, A1413302)
Geltrex LDEV-Free Reduced Growth Factor Basement Membrane Matrix (Cat. Nos. A1413201, A1413202) is the same as the product above. However, it has not been tested to verify it will support hESC growth and expansion. This is a "general" version and not specific to a cell type or line. A full product description can be found here:
Geltrex LDEV-Free Reduced Growth Factor Basement Membrane Matrix (Cat. Nos. A1413201, A1413202)
Geltrex LDEV-Free Reduced Growth Factor Basement Membrane Matrix (Cat. Nos. A1413201, A1413202)
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
How should I prepare Geltrex LDEV-Free, hESC-Qualified, Reduced Growth Factor Basement Membrane Matrix (Cat. No. A1413301, A1413302) stock solution, and how long is the stock solution stable after thawing?
We recommend thawing Geltrex LDEV-Free, hESC-Qualified, Reduced Growth Factor Basement Membrane Matrix (Cat. No. 1413301, 1413302) at 4 degrees C before making your stock solution. On ice, make a 1:1 dilution with DMEM/F-12 and aliquot into smaller volumes. Stock solution aliquots should be stored at -20 degrees C or used immediately. Once the stock solution is thawed, it should be used for coating plates immediately.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What is the difference between Geltrex LDEV-Free, hESC-Qualified, Reduced Growth Factor Basement Membrane Matrix and Geltrex hESC-Qualified, Ready-To-Use, Reduced Growth Factor Basement Membrane Matrix? I am working with human hepatocytes.
We do not have comparison data for the two Geltrex products. We recommend using the Geltrex LDEV-Free, hESC-Qualified, Reduced Growth Factor Basement Membrane Matrix for hepatocytes. We have not performed a significant amount of testing using the Geltrex hESC-Qualified, Ready-To-Use, Reduced Growth Factor Basement Membrane Matrix on these cells.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What substrate should I use for cardiomyocyte differentiation?
We recommend using Gibco Geltrex LDEV-Free, hESC Qualified Reduced Growth Factor Basement Membrane Matrix (Cat. No. A1413301 or A1413302) or Gibco Vitronectin (VTN-N) Recombinant Human Protein, Truncated (Cat. No. A14700) for xeno-free applications.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
How do I characterize human embryonic stem ( ES) cells?
Human ES cells are generally characterized by their typical morphology (they grow as tightly packed clusters of small cells with high ratio of nucleus to cytoplasm); surface marker expression; RT-PCR detection of stem cell-specific gene expression (such as Oct3/4, Sox2, and Nanog); alkaline phosphatase staining, and telomerase activity assay. The most commonly used ES specific surface markers include stage-specific embryonic antigens SSEA-3 and SSEA-4 for human ES cells. Other ES-specific surface antigens also include TRA-1-60 and TRA-1-81. (Science 282:1145 (1998).
How are human embryonic stem (ES) cells derived?
Human ES cells are derived from human blastocyst inner cell masses, isolated by immunosurgery with rabbit antiserum to BeWO cells (a human trophoblast cell line) (Science 282:1145 (1998)).
What are ES cells?
Embryonic stem (ES) cells are derived from the early mammalian embryo and are capable of unlimited, undifferentiated proliferation in vitro while maintaining their potential to differentiate into a wide of range of adult tissues including germ cells. The pluripotency of the ES cells is normally demonstrated in vitro by inducing ES cells to differentiate into embryoid bodies and checking lineage-specific markers for differentiated cells in three body layers (endo, meso, and ectoderm), or injecting them into immunodeficient mice and determining the cell types produced in the teratomas.
Can differentiation of cells occur when using Gibco Geltrex Matrix? Can this be prevented?
Yes, try a variety of dilutions to coat the plates. Some customers use a 1:100 dilution and have better results. The optimal dilution is cell-line dependent.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Why can Gibco Geltrex Matrix be used for adhesion-related research, as opposed to Gibco AlgiMatrix 3D Culture System which cannot?
Geltrex Matrix is a basement membrane extract that contains collagen IV, laminin, and other matrix proteins and growth factors that are required for cell adhesion. AlgiMatrix Matrix contains only alginate which does not facilitate cell adhesion. There is nothing in the alginate sponges for the cells to interact with.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Is it normal to see floating debris when pre-coating with Ready-to-Use Gibco Geltrex Matrix?
Yes, this is normal. It has been observed in product development. Remove the Gibco Geltrex Matrix solution after pre-coating and add cells as normal. H9 human embryonic stem cells have been cultured on dishes pre-coated with Ready-to-Use Gibco Geltrex Matrix with this observation in the pre-coating procedure.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What is LDEV?
Lactate dehydrogenase-elevating virus (LDEV) infects mouse macrophages and induces persistent infections in laboratory mice. LDEV, an RNA virus, alters in vivo physiology, and is a common contaminant of biological materials derived from transplantable tumor cell lines.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Can cells be removed from the Gibco Geltrex Matrix?
If you would like to culture the cells once recovered from the matrix, we recommend washing the cells first, then using trypsin/EDTA, TrypLE reagent, or a solution of 3,000 U/mL collagenase and 1,000 U hyaluronidase in low-glucose DMEM. If you would like to monitor cell surface markers, we recommend removal of the cells by mechanical means, such as a vigorous wash or a delicate scrape.
Note that the phenotype of the cells may change if they are cultured in 2D after their removal from the matrix.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Has Gibco Geltrex Matrix been tested with StemPRO hESC serum-free Medium?
Yes, these products were designed to work together.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What is Gibco Geltrex Matrix?
Gibco Geltrex Matrix is a soluble form of reduced growth factor (RGF) basement membrane extract (BME) purified from murine Engelbreth-Holm-Swarm tumor. Historically these preparations have contained the lactate dehydrogenase-elevating virus (LDEV), which does not impact the performance of the material but may be of concern in certain applications such as xenografts or other clinically focused research. New manufacturing methods are now available that allow for the removal of this virus. The resulting LDEV-free material is tested using PCR methods to confirm that the material is virus-free.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What are some general differences between Gibco Geltrex Matrix, collagen, AlgiMatrix 3D Culture System, and CTS Cellstart Substrate?
Gibco Geltrex Matrix and collagen (rat tail and bovine) can be used as either a coating solution or a 3D gel matrix. CELLstart Substrate was developed to be used as a xeno-free coating matrix for only ESC applications (as a substitute for Gibco Geltrex Matrix or Matrigel Matrix, which are of animal origin). AlgiMatrix Matrix is a 3D scaffold-type matrix that does not support cell attachment but does provide a good environment for growing spheroids that can be easily harvested for downstream applications.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.