Maleic acid, 98+%
Maleic acid, 98+%
Maleic acid, 98+%
Thermo Scientific Chemicals

Maleic acid, 98+%

CAS: 110-16-7 | C4H4O4 | 116.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
5000 g
Catalog number A14596.0B
also known as A14596-0B
Price (USD)
65.65
Special offer
Online exclusive
Ends: 28-Dec-2026
77.50
Save 11.85
Each
Quantity:
1000 g
Request bulk or custom format
Price (USD)
65.65
Special offer
Online exclusive
Ends: 28-Dec-2026
77.50
Save 11.85
Each
Chemical Identifiers
CAS110-16-7
IUPAC Name(2E)-but-2-enedioic acid
Molecular FormulaC4H4O4
InChI KeyVZCYOOQTPOCHFL-OWOJBTEDSA-N
SMILESOC(=O)\C=C\C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
FormCrystals or powder or crystalline powder
Appearance (Color)White
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%
Melting Point (clear melt)130-138?C
View more
Maleic acid is used as a precursor to fumaric acid, dimethyl maleate and glyoxalic acid. It is an electrophile and acts as dienophine in Diels-Alder reactions. It reacts with drugs to form more stable addition salts like indacaterol maleate, carfenazine, chlorpheniramine, pyrilamine, methylergonovine and thiethylperazine. Its maleate ion is useful in biochemistry as an inhibitor of transaminase reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Maleic acid is used as a precursor to fumaric acid, dimethyl maleate and glyoxalic acid. It is an electrophile and acts as dienophine in Diels-Alder reactions. It reacts with drugs to form more stable addition salts like indacaterol maleate, carfenazine, chlorpheniramine, pyrilamine, methylergonovine and thiethylperazine. Its maleate ion is useful in biochemistry as an inhibitor of transaminase reactions.

Solubility
Soluble in water, acetone, glacial acetic acid, alcohol and concentrated sulfuric acid. Slightly soluble in diethyl ether. Insoluble in benzene and chloroform.

Notes
Incompatible with oxidizing agents, bases, reducing agents, metals and alkalis.
RUO – Research Use Only

General References:

  1. Tsukigawa, K.; Liao, L.; Nakamura, H.; Fang, J.; Greish, K.; Otagiri, M.; Maeda, H. Synthesis and therapeutic effect of styrene-maleic acid copolymer-conjugated pirarubicin. Cancer Sci. 2015, 106 (3), 270-278.
  2. Filippov, A.; Tarabukina, E.; Simonova, M.; Kirila, T.; Fundueanu, G.; Harabagiu, V.; Constantin, M.; Popescu, I. Synthesis and Investigation of Double Stimuli-Responsive Behavior of N-Isopropylacrylamide and Maleic Acid Copolymer in Solutions. J. Macromol. Sci. Part B Phys. 2015, 54 (9), 1105-1121.