Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • CD16/CD32 Antibodies

          Invitrogen

          CD16/CD32 Monoclonal Antibody (93), PE-Cyanine7

          1 Reference
          View all (56) CD16/CD32 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite CD16/CD32 Monoclonal Antibody (93), PE-Cyanine7

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          CD16/CD32 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          CD16/CD32 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CD16/CD32 Antibody (A14719) in Flow

          Flow cytometry analysis of CD16/CD32 in BALB/c mouse splenocytes. Cells were stained using CD16/CD32 Monoclonal Antibody (93), PE-Cyanine7 (Product # A14719) and Rat Anti-Mouse CD19-PE. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CD16/CD32 Antibody in Flow Cytometry (Flow)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          A14719

          Applications
          Tested Dilution
          Publications

          Flow Cytometry (Flow)

          0.2 µg/1x10^6 cells
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Mouse

          Published species

          Mouse

          Host/Isotype

          Rat / IgG2a, lambda

          Class

          Monoclonal

          Type

          Antibody

          Clone

          93

          Immunogen

          Mouse CD16 and CD32 (common epitope).

          Conjugate

          PE-Cyanine7 PE-Cyanine7 PE-Cyanine7
          • FITC
          • Request custom conjugation

          Excitation/Emission Max

          569/780 nm View spectra spectra

          Form

          Liquid

          Concentration

          0.1 mg/mL

          Amount

          25 µg

          Purification

          purified

          Storage buffer

          PBS, pH 7.1, with proprietary stabilizer

          Contains

          <0.1% sodium azide

          Storage conditions

          4°C, store in dark, DO NOT FREEZE!

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2534235

          Target Information

          CD16 is a 50-65 kDa cell surface molecule that exists in two forms - a transmembranous form expressed by natural killer (NK) cells, and some T cells and a phosphatidylinositol linked form expressed by granulocytes. CD16 is a low affinity receptor for IgG (FcR III), and is an important receptor mediating ADCC (antibody dependent cell mediated cytotoxicity) by NK cells. IN comparison, CD32 is a 40 kD glycoprotein that acts as a low affinity receptor for IgG (also known as Fc gamma RII), and mediates several functions including endocytosis, activation of secretion, cytotoxicity and immunomodulation. CD32 is expressed by B cells, monocytes, granulocytes and platelets. Further, CD32 is involved in the phagocytosis of immune complexes, the regulation of antibody production by B-cells, and variations of the CD32 gene may increase vulnerability to systemic lupus erythematosus (SLE). Several transcript variants encoding different isoforms of CD32 have been found.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          How to use the Panel Builder

          Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

          Bioinformatics

          Protein Aliases: CD16A; CD16B; CD32 receptor 2; CDw32; fc gamma receptor IIB; Fc gamma receptor III; Fc gamma receptor precursor; Fc gamma RIIB; fc-gamma RII; Fc-gamma RII-b; Fc-gamma RII-c; fc-gamma RIII; Fc-gamma-1/gamma-2bR receptor precursor; Fc-gamma-R-alpha receptor; Fc-gamma-R-beta-1 precursor; Fc-gamma-R-beta-2 precursor; Fc-gamma-RII; fc-gamma-RIIB; Fc-gamma-RIIc; Fcg receptor III; FCG2; FcgammaRIIB; FcgammaRIII; FCGR2C; FCGR3A; FCGR3B; FCGRIII; FCR-10; FcRII-b; FcRII-c; fcRIII; FCRIIIA; FGFR2B; igG Fc receptor II beta; igG Fc receptor III; low affinity immunoglobulin gamma Fc region receptor II; lymphocyte antigen 17; RP11-5K23.1; unnamed protein product

          View more View less

          Gene Aliases: CD16; CD32; F630109E10Rik; Fc[g]RII; Fcgr2; Fcgr2a; FcgRII; Fcr-2; Fcr-3; fcRII; Ly-17; Ly-m20; LyM-1

          View more View less

          UniProt ID: (Mouse) P08101

          View more View less

          Entrez Gene ID: (Mouse) 14130, (Mouse) 14131

          View more View less

          Function(s)
          beta-amyloid binding protein binding IgG receptor activity low-affinity IgG receptor activity IgG binding immunoglobulin binding macromolecular complex binding immunoglobulin receptor binding
          Process(es)
          antibody-dependent cellular cytotoxicity negative regulation of type I hypersensitivity immune complex clearance negative regulation of immunoglobulin production negative regulation of inflammatory response to antigenic stimulus negative regulation of acute inflammatory response to antigenic stimulus negative regulation of humoral immune response mediated by circulating immunoglobulin receptor-mediated endocytosis phagocytosis, engulfment defense response cell surface receptor signaling pathway response to bacterium immunoglobulin mediated immune response dendrite development antigen processing and presentation of exogenous peptide antigen via MHC class II cerebellum development negative regulation of B cell proliferation endocytic recycling negative regulation of interleukin-10 production positive regulation of tumor necrosis factor production negative regulation of neutrophil apoptotic process Fc-gamma receptor signaling pathway positive regulation of JNK cascade regulation of long-term neuronal synaptic plasticity negative regulation of phagocytosis positive regulation of phagocytosis regulation of immune response negative regulation of immune response cellular response to molecule of bacterial origin Fc receptor-mediated immune complex endocytosis regulation of dendritic spine maintenance regulation of endoplasmic reticulum stress-induced neuron intrinsic apoptotic signaling pathway cellular response to beta-amyloid positive regulation of response to endoplasmic reticulum stress positive regulation of type IIa hypersensitivity positive regulation of type III hypersensitivity positive regulation of type I hypersensitivity serotonin secretion phagocytosis, recognition neutrophil chemotaxis antigen processing and presentation of exogenous peptide antigen via MHC class I mast cell activation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-76c4fffd86-jvpnf:80/100.66.74.151:80.
          git-commit: bdc067a1e9e92d056899a2dda64af631988aa8a1
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.54.0-Offline