Neurite Outgrowth Staining Kit - FAQs
Neurite Outgrowth Staining Kit - FAQs
View additional product information for Neurite Outgrowth Staining Kit - FAQs (A15001)
2 product FAQs found
Are the stains used in the Neurite Outgrowth Staining Kit specific for neurons?
No, the viability dye and membrane stain can stain any cell type. Other Neurite Outgrowth Staining Kit FAQs can be found here (https://www.thermofisher.com/content/dam/LifeTech/migration/files/cell-tissue-analysis/pdfs.par.90941.file.dat/neurite-outgrowth-staining-kit-faqs.pdf).
Do you offer any products to measure neuronal cell health?
PrestoBlue Cell Viability Stain and CyQUANT Cell Proliferation Assay Kit can be used. We also offer a Neurite Outgrowth Staining Kit (Cat. No. A15001). More information about our different assays for neuronal cell health can be found here (https://www.thermofisher.com/us/en/home/life-science/cell-analysis/neuroscience/neuronal-cell-health-assays.html).