Primary Mouse Cortical Neurons, 1 x 106 - FAQs
Primary Mouse Cortical Neurons, 1 x 106 - FAQs
View additional product information for Primary Mouse Cortical Neurons - FAQs (A15586, A15585)
3 product FAQs found
Would I be able to find out the sex of our Primary Mouse Cortical Neurons?
Primary Mouse Cortical Neurons are mixed sex, because it's too hard to identify the sex of a mouse fetus.
I'm currently culturing neurons. Which media system do you recommend using in conjunction with the Poly-D-Lysine matrix?
We recommend using the B-27 Plus Neuronal Culture System when culturing neurons on Poly-D-Lysine. The B-27 Plus Neuronal Culture System is a complete media system that includes the B-27 Plus Supplement and the Neurobasal Plus Medium.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What cell types will Poly-D-Lysine support as an attachment matrix?
The Poly-D-Lysine matrix has successfully supported the attachment of primary rodent neurons, iPSC-derived neurons and PC12 cells.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.