PolarScreen™ ER Alpha Competitor Assay, Red
Thermo Scientific™

PolarScreen™ ER Alpha Competitor Assay, Red

The PolarScreen™ ER (Estrogen Receptor) Alpha Competitor Assay Kit, Red, provides a sensitive and efficient method for high-throughput, fluorescence polarization-basedRead more
Have Questions?
Catalog NumberQuantity
A15884800 assays
Catalog number A15884
Price (USD)
2,325.00
Each
Quantity:
800 assays
Price (USD)
2,325.00
Each
The PolarScreen™ ER (Estrogen Receptor) Alpha Competitor Assay Kit, Red, provides a sensitive and efficient method for high-throughput, fluorescence polarization-based screening of potential ER alpha ligands. The kit uses insect cell-expressed, full-length, native (untagged) human estrogen receptor alpha (available separately, Cat. No. A15674) and a novel, high-affinity, fluorescent estrogen ligand (Fluormone EL Red) in a homogenous, mix-and-read assay format.

This kit produces a red color read-out; a 'green' kit is also available. Both kits suitably determine the IC50s of competitive compounds. Color choice depends on the spectral properties of the compounds to be screened and the available instrumentation. For additional information or assistance contact drugdiscoverytech@lifetech.com.

PolarScreen™ ER Alpha Competitor Assay, Red, is:

• Complete & ready-to-use out of the box—all reagents included, just add your test compounds
• High throughput compatible—800 x 20 μL fluorescence polarization assays in a 384-well plate
• Convenient—available in bulk sizes for larger screening needs

How the Assay Works
Full length ER alpha is added to a fluorescent estrogen ligand to form an ER-Fluormone EL complex. This complex is added to individual test compounds in multiwell plates. To determine relative affinity, the assay measures fluorescence polarization. Competing test compounds will displace the Fluormone EL ligand from ER alpha, permitting it to tumble rapidly and resulting in low polarization values. A shift in the fluorescence polarization value in the presence of test compound is used to determine the relative affinity of test compounds for ER alpha.

All components are manufactured under the strictest quality parameters. This product's performance, per the Certificate of Analysis, is guaranteed for 6 months after purchase. Contact us for bulk quantities or for assistance with other nuclear receptor assay needs, from assay development to library screening and beyond.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)Fluorescent Imager
Format384-well Plate
Quantity800 assays
Detection MethodFluorescence
TypeER Alpha Competitor Assay
Unit SizeEach
Contents & Storage
Contains:

• 1 tube full-length ER Alpha Recombinant Protein (store at -68 to -85°C)
• 1 tube of Fluormone EL Red Tracer (store at -5 to -30°C)
• 2 bottles of ER Red Screening Buffer (store at -5 to -30°C)

Frequently asked questions (FAQs)

How do I access the TaqMan hPSC Scorecard Analysis Software?

Please visit the website link.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Applications Support Center