N-Benzyloxycarbonylglycine, 98+%
N-Benzyloxycarbonylglycine, 98+%
N-Benzyloxycarbonylglycine, 98+%
Thermo Scientific Chemicals

N-Benzyloxycarbonylglycine, 98+%

CAS: 1138-80-3 | C10H11NO4 | 209.201 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A15928.14
also known as A15928-14
Price (USD)
40.65
Special offer
Online exclusive
Ends: 30-Jun-2026
47.60
Save 6.95 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
40.65
Special offer
Online exclusive
Ends: 30-Jun-2026
47.60
Save 6.95 (15%)
Each
Chemical Identifiers
CAS1138-80-3
IUPAC Name2-{[(benzyloxy)carbonyl]amino}acetic acid
Molecular FormulaC10H11NO4
InChI KeyCJUMAFVKTCBCJK-UHFFFAOYSA-N
SMILESOC(=O)CNC(=O)OCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%
Appearance (Color)White
N-Carbobenzoxyglycine is used in the dipeptide synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Carbobenzoxyglycine is used in the dipeptide synthesis.

Solubility
Soluble in methanol. Insoluble in water.

Notes
Store in cool dry place. Keep away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Robert M. Silverstein. The assay of the bromelains using Nα-CBZ-l-lysine p-nitrophenyl ester and N-CBZ-glycine p-nitrophenyl ester as substrates. Anal. Biochem. 1974, 62 (2),478-484 .
  2. Rosina López-Fandiño.; Iqbal Gill.; Evgeny N. Vulfson. Protease-catalyzed synthesis of oligopeptides in heterogenous substrate mixtures. Biotechnol. Bioeng. 1994, 43 (11),1024-1030 .