L-Lysine dihydrochloride, 99%
L-Lysine dihydrochloride, 99%
L-Lysine dihydrochloride, 99%
Thermo Scientific Chemicals

L-Lysine dihydrochloride, 99%

CAS: 657-26-1 | C6H16Cl2N2O2 | 219.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A16127.14
also known as A16127-14
Price (USD)
29.65
Online exclusive
33.10
Save 3.45
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
29.65
Online exclusive
33.10
Save 3.45
Each
Chemical Identifiers
CAS657-26-1
IUPAC Name(2S)-2,6-diaminohexanoic acid dihydrochloride
Molecular FormulaC6H16Cl2N2O2
InChI KeyJBBURJFZIMRPCZ-XRIGFGBMSA-N
SMILESCl.Cl.NCCCC[C@H](N)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder or crystalline powder
Water Content (Karl Fischer Titration)<3.50%
Optical Rotation13-17? (c=2, in water)
Appearance (Color)White to pale yellow
Assay (unspecified)≥98.5 to ≤101.5% (dry wt. basis)
View more
Synthesis of hyperbranched polylysine using the N-hydroxysuccinimide ester of L-lysine dihydrochloride as AB 2 building block. The histidine and lysine standards were prepared from L-histidine monohydro- chloride and L-lysine dihydrochloride respectively. L-Homocitrulline-The copper complex was prepared from L-lysine dihydrochloride in 80 to 86 per cent yields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Synthesis of hyperbranched polylysine using the N-hydroxysuccinimide ester of L-lysine dihydrochloride as AB 2 building block. The histidine and lysine standards were prepared from L-histidine monohydro- chloride and L-lysine dihydrochloride respectively. L-Homocitrulline-The copper complex was prepared from L-lysine dihydrochloride in 80 to 86 per cent yields.

Solubility
Soluble in water (100 mg/ml).

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.
RUO – Research Use Only

General References:

  1. A. Marcos-Fernández,; G.A. Abraham,; J.L. Valentín,; J. San Román. Synthesis and characterization of biodegradable non-toxic poly(ester-urethane-urea)s based on poly(ε-caprolactone) and amino acid derivatives . Polymer. 2006, 47(3), 785-798.
  2. Alton C. Kurtz. Use of copper (ii) ion in masking (y-amino groups of amino acids. The Journal of Biological Chemistry. 1949, 180, 1253-1267..