In order to support the growing demand for this product, an additional Thermo Fisher Scientific manufacturing site has begun producing these products. Please click here for more information »
Pair your qPCR Master Mix with ready to use TaqMan Gene Expression, Genotyping Assay or Custom Probes for multiplexing. Use our helpful TaqMan Search Wizard to find the Assays for your target of interest. Learn more
TaqPath™ qPCR Master Mix, CG
Applied Biosystems™

TaqPath™ qPCR Master Mix, CG

The master mix delivers confident results in both single and duplex reactions, even in the presence of inhibitors commonly foundRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A1624810 x 5 mL
A162452 x 5 mL
A152981 x 50 mL
A162475 x 5 mL
A152971 x 5 mL
Catalog number A16248
Price (USD)
6,262.50
Special offer
Online exclusive
Ends: 11-Dec-2026
8,350.00
Save 2,087.50
Each
Quantity:
10 x 5 mL
Price (USD)
6,262.50
Special offer
Online exclusive
Ends: 11-Dec-2026
8,350.00
Save 2,087.50
Each

The master mix delivers confident results in both single and duplex reactions, even in the presence of inhibitors commonly found in clinical samples. All lots are functionally tested to help ensure lot-to-lot reproducibility for Ct consistency and dynamic range across a wide variety of assays. This General Purpose Reagent is labeled 'For Laboratory Use' and manufactured in an ISO 13485-certified and FDA-registered facility that adheres to cGMP principles. With stringent quality and premier performance, TaqPath qPCR Master Mix is a superior choice for your diagnostic testing or development needs.

Features of the TaqPath qPCR Master Mix include:

  • Efficient and linear detection of up to eight logs with gene expression or miRNA assays
  • Enables reliable detection of low-copy templates with reproducible CT results
  • Robust multiplexing performance with exogenous or endogenous targets
  • Manufactured with stringent production and process controls to help ensure lot-to-lot consistency
  • Compliance Package containing most frequently requested compliance documents, available below

TaqPath qPCR Master Mix, CG, is a 2X formulation designed for gene expression and miRNA analysis. It contains thermostable Fast DNA polymerase, uracil-N-glycosylase (UNG), dNTPs with dUTP, ROX dye (passive reference), and optimized buffer components for maximum robustness and reproducibility.

Validated with a breadth of workflows

TaqPath qPCR Master Mix, CG, has been validated to provide high specificity and dynamic range for use in multiple real-time PCR applications. The formulation can be run in either fast or standard cycling conditions on a wide variety of qPCR platforms. The figure below demonstrates the excellent PCR linearity over an 8-log input range of template with both gene expression and miRNA assays.

In addition, TaqPath qPCR Master Mix, CG, has been engineered to retain consistent performance in preassembled reactions for up to 48 hours. The stability of this mix provides users of high-throughput, liquid handling systems the assurance that the results on the first plate will mimic those of the last plate. TaqPath qPCR Master Mix has been benchmarked against similar competitor master mixes and demonstrates equivalent or better sensitivity and dynamic range across a variety of targets.

Reproducible sensitive detection

We understand the importance of reliably detecting low-copy targets for your test quality and data interpretation. TaqPath qPCR Master Mix, CG, helps generate significant and reproducible CT values for ≤10 copy detection. The figure below shows the consistent CT results obtained from three unique lots when detecting 10-copy inputs. This lot-to-lot CT consistency is preserved across multiple assays with different attributes and expression levels to maximize confidence in your results.

Optimized for multiplexing

Simultaneous amplification of multiple assays can be beneficial not only as a control to normalize and detect anomalies in experiments, but also to improve efficiency and cost-savings for labs. TaqPath qPCR Master Mix has been optimized with enzymes and component concentrations to facilitate multiplexing while preserving specificity, and it is validated for duplex performance in each manufacturing lot.

Inhibitor tolerant

Unlike other master mixes on the market, TaqPath qPCR Master Mix's unique proprietary formulation allows robust performance even in the presence of substances that can normally inhibit PCR, such as heparin, hematin, or EDTA, increasing your confidence when working with a variety of complex clinical samples. TaqPath master mixes demonstrate increased tolerance to inhibitors compared to competitor mixes.

Compliance Package

In an effort to efficiently support your compliance needs, frequently requested compliance documents are available here:

  • Statement of Quality relating to the current good manufacturing practices (cGMP) to which TaqPath master mixes adhere
  • Detailed lot-specific Certificate of Analysis (COA) (see below in Documents & Downloads section)
  • Supplier Evaluation Support
  • For EU Reach compliance-related inquiries, email LSGChemicalCompliance@thermofisher.com
  • BSE/TSE Statement
  • Change Control and Notification Registration

Not finding the compliance document you're looking for? Technical Support is here to help.

Specifications
For Use With (Equipment)QuantStudio Dx, 7500 Fast Dx, Roche LightCycler PRO System, Bio-Rad CFX Opus 96 Real-Time PCR System
FormatBottle
Hot StartBuilt-In Hot Start
IncludesdUTP, UNG/UDG
Inhibitor Tolerance (Simple)High
Multiplex CapabilityDuplex
No. of Reactions5000 Reactions
Passive Reference DyeROX (Pre-mixed)
PolymeraseFast DNA Polymerase
Product LineTaqPath
Product TypeqPCR Master Mix
Quantity10 x 5 mL
Sample TypecDNA, DNA
SensitivityDown to 10 copies
Sufficient For5000 Reactions
Target SpecificitycDNA or gDNA only
Thermostability48 hr., Assembled Reaction
Volume10 x 5 mL
Concentration2X
Detection MethodPrimer-probe
For Use With (Application)Gene Expression, Microbial Detection, DNA Quantification, Presence/Absence, miRNA analysis
FormLiquid
PCR MethodqPCR
Reaction SpeedFast or Standard
Unit SizeEach
Contents & Storage
Store at 2°C to 8°C in the dark.