DL-Valine, 99%
DL-Valine, 99%
DL-Valine, 99%
Thermo Scientific Chemicals

DL-Valine, 99%

CAS: 516-06-3 | C5H11NO2 | 117.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A16756.0E
also known as A16756-0E
Price (USD)
579.65
Online exclusive
644.00
Save 64.35
Each
Quantity:
2500 g
Request bulk or custom format
Price (USD)
579.65
Online exclusive
644.00
Save 64.35
Each
Chemical Identifiers
CAS516-06-3
IUPAC Name2-amino-3-methylbutanoic acid
Molecular FormulaC5H11NO2
InChI KeyKZSNJWFQEVHDMF-UHFFFAOYSA-N
SMILESCC(C)C(N)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder or crystalline powder or flakes
Assay (Non-aqueous acid-base Titration)≥98.5 to ≤101.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only