Thermo Scientific Chemicals

5-Aminolevulinic acid hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: A16942.04
2 g, Each
Thermo Scientific Chemicals

5-Aminolevulinic acid hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: A16942.04
2 g, Each
Quantity
Catalog number: A16942.04
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5451-09-2
IUPAC Name
hydrogen 5-amino-4-oxopentanoic acid chloride
Molecular Formula
C5H10ClNO3
InChI Key
ZLHFONARZHCSET-UHFFFAOYSA-N
SMILES
[H+].[Cl-].NCC(=O)CCC(O)=O
Form
Crystals or powder or crystalline powder
Comment
Material Sourced in UK and US
Assay (Titration ex Chloride)
≥98.5 to ≤101.5% (UK Sourced Material)
Identification (FTIR)
Conforms (UK Sourced Material)
Appearance (Color)
White to cream

Description

5-Aminolevulinic acid hydrochloride finds an important role as a precursor in the synthesis of tetrapyrroles such as chlorophyll and heme. It is widely utilized in photodynamic therapy of diseases namely, Paget’s disease and human papillomavirus (HPV) infection-associated cervical condylomata acuminata.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in dimethyl sulfoxide, methanol and water.

Notes
Store in a cool place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text