beta-Alanine ethyl ester hydrochloride, 98%
beta-Alanine ethyl ester hydrochloride, 98%
beta-Alanine ethyl ester hydrochloride, 98%
Thermo Scientific Chemicals

beta-Alanine ethyl ester hydrochloride, 98%

CAS: 4244-84-2 | C5H12ClNO2 | 153.61 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number A17277.09
also known as A17277-09
Price (USD)
54.65
Online Exclusive
60.60
Save 5.95 (10%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
54.65
Online Exclusive
60.60
Save 5.95 (10%)
Each
Chemical Identifiers
CAS4244-84-2
IUPAC Nameethyl 3-aminopropanoate hydrochloride
Molecular FormulaC5H12ClNO2
InChI KeyRJCGNNHKSNIUAT-UHFFFAOYSA-N
SMILESCl.CCOC(=O)CCN
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystalline and/or lumpy powder
CommentMay contain up to ca. 2% water
Assay (Titration ex Chloride)≥97.5 to ≤102.5%
Melting Point (clear melt)59-72?C
Appearance (Color)White to pale cream
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only