FluoroBrite™ DMEM, 500 mL - FAQs
FluoroBrite™ DMEM, 500 mL - FAQs
View additional product information for FluoroBrite™ DMEM - FAQs (A1896701, A1896702)
3 product FAQs found
What is the osmolality of Fluorobrite DMEM?
We do provide osmolality information on the certificate of analysis. All lots of Fluorobrite DMEM (Cat. Nos. A1896701 and A1896702) will meet the osmolality specification of 320-350 mOsm/kg.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
I understand that some media are worse than others for fluorescence imaging. How do I choose?
Most media contain phenol red, which can quench fluorescent dyes in the visible wavelengths. Most media also contain autofluorescent components, such as riboflavin, which can reduce signal-to-background. We offer FluoroBrite DMEM and HEPES-based Live Cell Imaging Solution, which have been optimized for fluorescent imaging. We also offer a number of media without phenol red. But if none of these are reasonable options for your experiment, then we also offer BackDrop Background Suppressor ReadyProbes Reagent, which can be added to quench media autofluorescence.
Should I be concerned about phenol red in my media when labeling my live cells with fluorescent dyes?
Some cell types accumulate phenol red, and this can pose a problem in the use of many fluorescent probes. Phenol red can quench visible-wavelength dyes and, although phenol red is non-fluorescent, various impurities may be fluorescent. We have many phenol red-free media to choose from. Our Live Cell Imaging Solution (HEPES-based) and our FluoroBrite DMEM have been optimized to be phenol red-free as well as to be non-autofluorescent.