Thermo Scientific Chemicals

L-(+)-2-Phenylglycine, 98+%

Catalog number: A19360.14
25 g, Each
Thermo Scientific Chemicals

L-(+)-2-Phenylglycine, 98+%

Catalog number: A19360.14
25 g, Each
Quantity
Catalog number: A19360.14
also known as A19360-14
Price (USD)
Quantity
-

Chemical Identifiers

CAS
2935-35-5
IUPAC Name
2-amino-2-phenylacetic acid
Molecular Formula
C8H9NO2
InChI Key
ZGUNAGUHMKGQNY-UHFFFAOYNA-N
SMILES
NC(C(O)=O)C1=CC=CC=C1
Appearance (Color)
White
Form
Crystals or powder or crystalline powder
Optical Rotation
157.0 ± 2.0? (C=1 in 1N HCl)
Assay (Non-aqueous acid-base Titration)
≥98.0 to ≤102.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text