Get upto 200X more sensitivity in ICC/IHC/FISH for more clarity and definition with new Tyramide SuperBoost™ kits
Alexa Fluor™ 532 NHS Ester (Succinimidyl Ester)
Invitrogen™

Alexa Fluor™ 532 NHS Ester (Succinimidyl Ester)

Alexa Fluor™ 532 is a bright yellow dye with excitation ideally suited for the frequency-doubled Nd:YAG laser line. Used forRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A20001
also known as A-20001
1 mg
A20101MP
also known as A-20101MP
5 mg
Catalog number A20001
also known as A-20001
Price (USD)
495.00
Each
Quantity:
1 mg
Request bulk or custom format
Price (USD)
495.00
Each

Alexa Fluor™ 532 is a bright yellow dye with excitation ideally suited for the frequency-doubled Nd:YAG laser line. Used for stable signal generation in imaging and flow cytometry, Alexa Fluor™ 532 dye is water soluble and pH-insensitive from pH 4 to pH 10. In addition to reactive dye formulations, we offer Alexa Fluor™ 532 dye conjugated to a variety of antibodies, peptides, proteins, tracers, and amplification substrates optimized for cellular labeling and detection (learn more).

The NHS ester (or succinimidyl ester) of Alexa Fluor™ 532 is the most popular tool for conjugating this dye to a protein or antibody. NHS esters can be used to label to the primary amines (R-NH2) of proteins, amine-modified oligonucleotides, and other amine-containing molecules. The resulting Alexa Fluor™ conjugate will exhibit brighter fluorescence and greater photostability than the conjugates of other spectrally similar fluorophores.

Detailed information about this AlexaFluor™ NHS ester:

  • Fluorophore label: Alexa Fluor™ 532 dye
  • Reactive group: NHS ester
  • Reactivity: Primary amines on proteins and ligands, amine-modified oligonucleotides
  • Ex/Em of the conjugate: 530/555 nm
  • Extinction coefficient: 81,000 cm-1M-1
  • Molecular weight: 723.8

Typical Conjugation Reaction

You can conjugate amine-reactive reagents with virtually any protein or peptide (the provided protocol is optimized for IgG antibodies). You can scale the reaction for any amount of protein, but the concentration of the protein should be at least 2 mg/mL for optimal results. We recommend trying three different degrees of labeling, using three different molar ratios of the reactive reagent to protein.

The Alexa Fluor™ NHS ester is typically dissolved in high-quality anhydrous dimethylformamide (DMF) or dimethylsulfoxide (DMSO) (D12345), and the reaction is carried out in 0.1–0.2 M sodium bicarbonate buffer, pH 8.3, at room temperature for 1 hour. Because the pKa of the terminal amine is lower than that of the lysine epsilon-amino group, you may achieve more selective labeling of the amine terminus using a buffer closer to neutral pH.

Conjugate Purification

Labeled antibodies are typically separated from free Alexa Fluor™ dye using a gel filtration column, such as Sephadex™ G-25, BioGel™ P-30, or equivalent. For much larger or smaller proteins, select a gel filtration media with an appropriate molecular weight cut-off or purify by dialysis. We offer several purification kits optimized for different quantities of antibody conjugate:

  • Antibody Conjugate Purification Kit for 0.5-1 mg (A33086)
  • Antibody Conjugate Purification Kit for 20-50 μg (A33087)
  • Antibody Conjugate Purification kit for 50-100 μg (A33088)

Learn More About Protein and Antibody Labeling

We offer a wide selection of Molecular Probes™ antibody and protein labeling kits to fit your starting material and your experimental setup. See our Antibody Labeling kits or use our Labeling Chemistry Selection Tool for other choices. To learn more about our labeling kits, read Kits for Labeling Proteins and Nucleic Acids—Section 1.2 in The Molecular Probes™ Handbook.

We'll Make a Custom Conjugate for You

If you can't find what you're looking for in our online catalog, we'll prepare a custom antibody or protein conjugate for you. Our custom conjugation service is efficient and confidential, and we stand by the quality of our work. We are ISO 13485:2000 certified.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Emission557 nm
For Use With (Application)Labeling chemistry, protein labeling and crosslinking
Label or DyeAlexa Fluor™ 532
Molecular Weight (g/mol)723.8 kDa
Product TypeAlexa Fluor™ 532 NHS Ester (Succinimidyl Ester)
PurityHPLC, 82% (at 254 nm)
Quantity1 mg
ColorRed
pH Range4 to 10
Unit SizeEach
Contents & Storage
Store at -5°C to -30°C.

Frequently asked questions (FAQs)

I am labeling a protein with Alexa Fluor 488 SDP ester. The manual recommends using a sodium bicarbonate buffer at pH 8.3. Can I use a different buffer instead?

Yes. The important thing is to use a buffered solution with a pH between 8.0 and 8.5. Do not use Tris buffer, which has amine groups. Most other buffers will work fine in that pH range. This is also true for other amine-reactive dyes, such as succinimidyl (NHS) esters or TFP esters.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

I am not going to use all of my Alexa Fluor succinimidyl ester reactive dye. Can I just make it up in DMSO and store aliquots at -20 degrees C?

This is not recommended. Any trace amounts of water in the DMSO can promote spontaneous hydrolysis over time. Even if using anhydrous DMSO, DMSO is hygroscopic; it readily absorbs moisture from the atmosphere over time. A better alternative is to dissolve the reactive dye in a volatile solvent, make smaller aliquots and then evaporate off the solvent using a vacuum pump. The smaller aliquots of solid reactive dye should then be stored frozen, desiccated and protected from light. Contact Technical Support by sending an email to techsupport@thermofisher.com for the recommended volatile solvent.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Citations & References (23)

Citations & References
Abstract
Alexa dyes, a series of new fluorescent dyes that yield exceptionally bright, photostable conjugates.
Authors:Panchuk-Voloshina N, Haugland RP, Bishop-Stewart J, Bhalgat MK, Millard PJ, Mao F, Leung WY, Haugland RP
Journal:J Histochem Cytochem
PubMed ID:10449539
'Alexa 350, Alexa 430, Alexa 488, Alexa 532, Alexa 546, Alexa 568, and Alexa 594 dyes are a new series of fluorescent dyes with emission/excitation spectra similar to those of AMCA, Lucifer Yellow, fluorescein, rhodamine 6G, tetramethylrhodamine or Cy3, lissamine rhodamine B, and Texas Red, respectively (the numbers in the ... More
Benzodiazepine binding studies on living cells: application of small ligands for fluorescence correlation spectroscopy.
Authors:Hegener O, Jordan R, Häberlein H
Journal:Biol Chem
PubMed ID:12530545
'We demonstrate the applicability of fluorescence correlation spectroscopy (FCS) for receptor binding studies using low molecular weight ligands on the membranes of living nerve cells. The binding of the benzodiazepine Ro 7-1986/602 (N-des-diethyl-fluorazepam), labeled with the fluorophore Alexa 532, to the benzodiazepine receptor was analyzed quantitatively at the membrane of ... More
Lateral mobility and specific binding to GABA(A) receptors on hippocampal neurons monitored by fluorescence correlation spectroscopy.
Authors:Meissner O, Häberlein H
Journal:Biochemistry
PubMed ID:12578381
'The binding behavior of a fluorescently labeled muscimol derivative to the GABA(A) receptor was analyzed at rat hippocampal neurons by fluorescence correlation spectroscopy. After muscimol had been labeled with the fluorophore Alexa Fluor 532, specific binding constants for binding of the dye-labeled ligand (Mu-Alexa) to the GABA(A) receptor were determined. ... More
Dynamics of beta2-adrenergic receptor-ligand complexes on living cells.
Authors:Hegener O, Prenner L, Runkel F, Baader SL, Kappler J, Häberlein H
Journal:Biochemistry
PubMed ID:15147203
'The agonist-induced dynamic regulation of the beta(2)-adrenergic receptor (beta(2)-AR) on living cells was examined by means of fluorescence correlation spectroscopy (FCS) using a fluorescence-labeled arterenol derivative (Alexa-NA) in hippocampal neurons and in alveolar epithelial type II cell line A549. Alexa-NA specifically bound to the beta(2)-AR of neurons with a K(D) ... More
Quantitative screening of single copies of human papilloma viral DNA without amplification.
Authors:Li J, Lee JY, Yeung ES
Journal:Anal Chem
PubMed ID:16970325
'We describe a novel quantitative viral screening method based on single-molecule detection that does not require amplification. DNA of human papilloma virus (HPV), the major etiological agent of cervical cancer, served as the screening target in this study. Eight 100-nucleotide single-stranded DNA probes were designed complementary to the E6-E7 gene ... More