Alexa Fluor™ 555 alkyne, Triethylammonium Salt
Invitrogen™

Alexa Fluor™ 555 alkyne, Triethylammonium Salt

The orange-fluorescent Alexa Fluor® 555 alkyne is designed to react with azide-containing molecules via the fast, selective and extremely efficientRead more
Have Questions?
Catalog NumberQuantity
A200130.5 mg
Catalog number A20013
Price (USD)
660.00
Each
Quantity:
0.5 mg
Request bulk or custom format
Price (USD)
660.00
Each
The orange-fluorescent Alexa Fluor® 555 alkyne is designed to react with azide-containing molecules via the fast, selective and extremely efficient copper-catalyzed “click" reaction.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical ReactivityAzide
Detection MethodFluorescence
Emission565
Excitation555
Label or DyeAlexa Fluor™ 555
Product TypeAlkyne
Quantity0.5 mg
Reactive MoietyAlkyne
Shipping ConditionRoom Temperature
System TypeClick-iT™
ColorOrange
Label TypeAlexa Fluor
Product LineAlexa Fluor
Unit SizeEach
Contents & Storage
Store at -20°C, dessicate and protect from light.

Citations & References (1)

Citations & References
Abstract
Chlamydia trachomatis inclusions induce asymmetric cleavage furrow formation and ingression failure in host cells.
Authors:Sun HS, Wilde A, Harrison RE,
Journal:Mol Cell Biol
PubMed ID:21969606
'Chlamydia trachomatis infection has been suggested to induce host genome duplication and is linked to increased risks of cervical cancer. We describe here the mechanism by which Chlamydia causes a cleavage furrow defect that consistently results in the formation of multinucleated host cells, a phenomenon linked to tumorigenesis. Host signaling ... More