Alexa Fluor™ 488 NHS Ester (Succinimidyl Ester), 5 mg - FAQs
View additional product information for Alexa Fluor™ 488 NHS Ester (Succinimidyl Ester) - FAQs (A20100, A20000)
Error:This product does not have any FAQs available.