Alexa Fluor™ 546 NHS Ester (Succinimidyl Ester)
Invitrogen™

Alexa Fluor™ 546 NHS Ester (Succinimidyl Ester)

Alexa Fluor™ 546 is a bright yellow dye. Used in imaging and flow cytometry, Alexa Fluor™ 546 dye is waterRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A201025 mg
A200021 mg
Catalog number A20102
Price (MXN)
-
Quantity:
5 mg

Alexa Fluor™ 546 is a bright yellow dye. Used in imaging and flow cytometry, Alexa Fluor™ 546 dye is water soluble and pH-insensitive from pH 4 to pH 10. In addition to reactive dye formulations, we offer Alexa Fluor™ 546 dye conjugated to a variety of antibodies, peptides, proteins, tracers, and amplification substrates optimized for cellular labeling and detection (learn more).

The NHS ester (or succinimidyl ester) of Alexa Fluor™ 546 is the most popular tool for conjugating this dye to a protein or antibody. NHS esters can be used to label to the primary amines (R-NH2) of proteins, amine-modified oligonucleotides, and other amine-containing molecules. The resulting Alexa Fluor™ conjugate will exhibit brighter fluorescence and greater photostability than the conjugates of other spectrally similar fluorophores.

Detailed information about this AlexaFluor™ NHS ester:
Fluorophore label: Alexa Fluor™ 546 dye
Reactive group: NHS ester
Reactivity: Primary amines on proteins and ligands, amine-modified oligonucleotides
Ex/Em of the conjugate: 554/570 nm
Extinction coefficient: 112,000 cm-1M-1
Molecular weight: 1159.6

Typical Conjugation Reaction
You can conjugate amine-reactive reagents with virtually any protein or peptide (the provided protocol is optimized for IgG antibodies). You can scale the reaction for any amount of protein, but the concentration of the protein should be at least 2 mg/mL for optimal results. We recommend trying three different degrees of labeling, using three different molar ratios of the reactive reagent to protein.

The Alexa Fluor™ NHS ester is typically dissolved in high-quality anhydrous dimethylformamide (DMF) or dimethylsulfoxide (DMSO) (D12345), and the reaction is carried out in 0.1–0.2 M sodium bicarbonate buffer, pH 8.3, at room temperature for 1 hour. Because the pKa of the terminal amine is lower than that of the lysine epsilon-amino group, you may achieve more selective labeling of the amine terminus using a buffer closer to neutral pH.

Conjugate Purification
Labeled antibodies are typically separated from free Alexa Fluor™ dye using a gel filtration column, such as Sephadex™ G-25, BioGel™ P-30, or equivalent. For much larger or smaller proteins, select a gel filtration media with an appropriate molecular weight cut-off or purify by dialysis. We offer several purification kits optimized for different quantities of antibody conjugate:
Antibody Conjugate Purification Kit for 0.5-1 mg (A33086)
Antibody Conjugate Purification Kit for 20-50 μg (A33087)
Antibody Conjugate Purification kit for 50-100 μg (A33088)

Learn More About Protein and Antibody Labeling
We offer a wide selection of Molecular Probes™ antibody and protein labeling kits to fit your starting material and your experimental setup. See our Antibody Labeling kits or use our Labeling Chemistry Selection Tool for other choices. To learn more about our labeling kits, read Kits for Labeling Proteins and Nucleic Acids—Section 1.2 in The Molecular Probes™ Handbook.

We’ll Make a Custom Conjugate for You
If you can’t find what you’re looking for in our online catalog, we’ll prepare a custom antibody or protein conjugate for you. Our custom conjugation service is efficient and confidential, and we stand by the quality of our work. We are ISO 13485:2000 certified.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical ReactivityAmine
Emission570 nm
Excitation554 nm
Label or DyeAlexa Fluor™ 546
Product TypeDye
Quantity5 mg
Reactive MoietyActive Ester, Succinimidyl Ester
Shipping ConditionRoom Temperature
Label TypeAlexa Fluor
Product LineAlexa Fluor
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -30°C) and protect from light.

Frequently asked questions (FAQs)

I am labeling a protein with Alexa Fluor 488 SDP ester. The manual recommends using a sodium bicarbonate buffer at pH 8.3. Can I use a different buffer instead?

Yes. The important thing is to use a buffered solution with a pH between 8.0 and 8.5. Do not use Tris buffer, which has amine groups. Most other buffers will work fine in that pH range. This is also true for other amine-reactive dyes, such as succinimidyl (NHS) esters or TFP esters.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

I am not going to use all of my Alexa Fluor succinimidyl ester reactive dye. Can I just make it up in DMSO and store aliquots at -20 degrees C?

This is not recommended. Any trace amounts of water in the DMSO can promote spontaneous hydrolysis over time. Even if using anhydrous DMSO, DMSO is hygroscopic; it readily absorbs moisture from the atmosphere over time. A better alternative is to dissolve the reactive dye in a volatile solvent, make smaller aliquots and then evaporate off the solvent using a vacuum pump. The smaller aliquots of solid reactive dye should then be stored frozen, desiccated and protected from light. Contact Technical Support by sending an email to techsupport@thermofisher.com for the recommended volatile solvent.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Citations & References (38)

Citations & References
Abstract
A small-molecule approach to studying invasive mechanisms of Toxoplasma gondii.
Authors:Carey KL, Westwood NJ, Mitchison TJ, Ward GE
Journal:Proc Natl Acad Sci U S A
PubMed ID:15123807
'Toxoplasma gondii is the most common protozoan parasite of humans. Infection with T. gondii can lead to life-threatening disease as a result of repeated cycles of host cell invasion, parasite replication, and host cell lysis. Relatively little is known about the invasive mechanisms of T. gondii and related parasites within ... More
CD24 is expressed by myofiber synaptic nuclei and regulates synaptic transmission.
Authors:Jevsek M, Jaworski A, Polo-Parada L, Kim N, Fan J, Landmesser LT, Burden SJ
Journal:Proc Natl Acad Sci U S A
PubMed ID:16606832
'The genes encoding several synaptic proteins, including acetylcholine receptors, acetylcholinesterase, and the muscle-specific kinase, MuSK, are expressed selectively by a small number of myofiber nuclei positioned near the synaptic site. Genetic analysis of mutant mice suggests that additional genes, expressed selectively by synaptic nuclei, might encode muscle-derived retrograde signals that ... More
Optimizing antibody immobilization strategies for the construction of protein microarrays.
Authors:Peluso P, Wilson DS, Do D, Tran H, Venkatasubbaiah M, Quincy D, Heidecker B, Poindexter K, Tolani N, Phelan M, Witte K, Jung LS, Wagner P, Nock S
Journal:Anal Biochem
PubMed ID:12531195
'Antibody microarrays have the potential to revolutionize protein expression profiling. The intensity of specific signal produced on a feature of such an array is related to the amount of analyte that is captured from the biological mixture by the immobilized antibody (the "capture agent"). This in turn is a function ... More
Temporally resolved interactions between antigen-stimulated IgE receptors and Lyn kinase on living cells.
Authors:Larson DR, Gosse JA, Holowka DA, Baird BA, Webb WW
Journal:J Cell Biol
PubMed ID:16275755
'Upon cross-linking by antigen, the high affinity receptor for immunoglobulin E (IgE), FcepsilonRI, is phosphorylated by the Src family tyrosine kinase Lyn to initiate mast cell signaling, leading to degranulation. Using fluorescence correlation spectroscopy (FCS), we observe stimulation-dependent associations between fluorescently labeled IgE-FcepsilonRI and Lyn-EGFP on individual cells. We also ... More
N-formyl peptide receptor phosphorylation domains differentially regulate arrestin and agonist affinity.
Authors:Key TA, Foutz TD, Gurevich VV, Sklar LA, Prossnitz ER
Journal:J Biol Chem
PubMed ID:12424254
'Arrestins regulate the signaling and endocytosis of many G protein-coupled receptors (GPCRs). It has been suggested that the functions of arrestins are dependent upon both the number and pattern of phosphorylation sites present in an activated GPCR. However, little is currently known about the relationships between the sites of receptor ... More