Alexa Fluor&trade; 647 C<sub>2</sub> Maleimide
Alexa Fluor&trade; 647 C<sub>2</sub> Maleimide
Alexa Fluor&trade; 647 C<sub>2</sub> Maleimide
Alexa Fluor&trade; 647 C<sub>2</sub> Maleimide
Invitrogen™

Alexa Fluor™ 647 C2 Maleimide

Alexa Fluor™ 647 is a bright, far-red fluorescent dye with excitation ideally suited for the 594 nm or 633 nmRead more
Have Questions?
Catalog NumberQuantity
A203471 mg
Catalog number A20347
Price (USD)
458.00
Each
Quantity:
1 mg
Request bulk or custom format
Price (USD)
458.00
Each
Alexa Fluor™ 647 is a bright, far-red fluorescent dye with excitation ideally suited for the 594 nm or 633 nm laser lines. Used for stable signal generation in imaging and flow cytometry, Alexa Fluor™ 647 dye is water soluble and pH-insensitive from pH 4 to pH 10. In addition to reactive dye formulations, we offer Alexa Fluor™ 647 dye conjugated to a variety of antibodies, peptides, proteins, tracers, and amplification substrates optimized for cellular labeling and detection (learn more).

The maleimide derivative of Alexa Fluor™ 647 is the most popular tool for conjugating the dye to a thiol group on a protein, oligonucleotide thiophosphate, or low molecular weight ligand. The resulting Alexa Fluor™ 647 conjugates exhibit brighter fluorescence and greater photostability than the conjugates of other spectrally similar fluorophores.

Detailed information about this AlexaFluor™ maleimide:

Fluorophore label: Alexa Fluor™ 647 dye
Reactive group: maleimide
Reactivity: thiol groups on proteins and ligands, oligonucleotide thiophosphates
Ex/Em of the conjugate: 651/671 nm
Extinction coefficient: 265,000 cm-1M-1
Spectrally similar dyes: Cy5
Molecular weight: ∼1250

Typical Conjugation Reaction
The protein should be dissolved at a concentration of 50-100 μM in a suitable buffer (10-100 mM phosphate, Tris, or HEPES) at pH 7.0-7.5. In this pH range, the protein thiol groups are sufficiently nucleophilic that they react almost exclusively with the reagent in the presence of the more numerous protein amine groups, which are protonated and relatively unreactive. We recommend reducing any disulfide bonds at this point using a 10-fold molar excess of reducing agent such as DTT or TCEP. Excess DTT must be removed by dialysis and subsequent thiol-modification should be carried out under oxygen-free conditions to prevent reformation of the disulfide bonds; these precautions are not necessary when using TCEP prior to maleimide conjugation.

The Alexa Fluor™ maleimide is typically dissolved in high-quality anhydrous dimethylsulfoxide (DMSO) at a concentration of 1-10 mM immediately prior to use, and stock solutions should be protected from light as much as possible. Generally, this stock solution is added to the protein solution dropwise while stirring to produce approximately 10-20 moles of reagent per mole of protein, and the reaction is allowed to proceed at room temperature for 2 hours or at 4°C overnight, protected from light. Any unreacted thiol-reactive reagent can be consumed by adding excess glutathione, mercaptoethanol, or other soluble low molecular weight thiol.

Conjugate Purification
Labeled antibodies are typically separated from free Alexa Fluor™ dye using a gel filtration column, such as Sephadex™ G-25, BioGel™ P-30, or equivalent. For much larger or smaller proteins, select a gel filtration media with an appropriate molecular weight cut-off or purify by dialysis. We offer several purification kits optimized for different quantities of antibody conjugate:
Antibody Conjugate Purification Kit for 0.5-1 mg (A33086)
Antibody Conjugate Purification Kit for 20-50 μg (A33087)
Antibody Conjugate Purification kit for 50-100 μg (A33088)

Learn More About Protein and Antibody Labeling
We offer a wide selection of Molecular Probes™ antibody and protein labeling kits to fit your starting material and your experimental setup. See our Antibody Labeling kits or use our Labeling Chemistry Selection Tool for other choices. To learn more about our labeling kits, read Kits for Labeling Proteins and Nucleic Acids—Section 1.2 in The Molecular Probes™ Handbook.

We’ll Make a Custom Conjugate for You
If you can’t find what you’re looking for in our online catalog, we’ll prepare a custom antibody or protein conjugate for you. Our custom conjugation service is efficient and confidential, and we stand by the quality of our work. We are ISO 13485:2000 certified.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical ReactivityThiol
Emission671 nm
Excitation651 nm
Label or DyeAlexa Fluor™ 647
Product TypeDye
Quantity1 mg
Reactive MoietyMaleimide
Shipping ConditionRoom Temperature
Label TypeAlexa Fluor
Product LineAlexa Fluor
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -30°C) and protect from light.

Citations & References (39)

Citations & References
Abstract
Membrane protein stoichiometry determined from the step-wise photobleaching of dye-labelled subunits.
Authors:Das SK, Darshi M, Cheley S, Wallace MI, Bayley H
Journal:Chembiochem
PubMed ID:17503420
Reversible transition between the surface trimer and membrane-inserted monomer of annexin 12.
Authors:Ladokhin AS, Haigler HT
Journal:Biochemistry
PubMed ID:15736950
'Under mildly acidic conditions, annexin 12 (ANX) inserts into lipid membranes to form a transbilayer pore [Langen, R., et al. (1998) Proc. Natl. Acad. Sci. U.S.A. 95, 14060]. In this study, we have addressed the question of the oligomeric state of ANX in this transbilayer conformation by means of Forster-type ... More
Single-molecule studies of synaptotagmin and complexin binding to the SNARE complex.
Authors:Bowen ME, Weninger K, Ernst J, Chu S, Brunger AT
Journal:Biophys J
PubMed ID:15821166
'The assembly of multiprotein complexes at the membrane interface governs many signaling processes in cells. However, very few methods exist for obtaining biophysical information about protein complex formation at the membrane. We used single molecule fluorescence resonance energy transfer to study complexin and synaptotagmin interactions with the SNARE complex in ... More
Chaperoning of insertion of membrane proteins into lipid bilayers by hemifluorinated surfactants: application to diphtheria toxin.
Authors:Palchevskyy SS, Posokhov YO, Olivier B, Popot JL, Pucci B, Ladokhin AS
Journal:Biochemistry
PubMed ID:16489756
'Hemifluorinated compounds, such as HF-TAC, make up a novel class of nondetergent surfactants designed to keep membrane proteins soluble under nondissociating conditions [Breyton, C., et al. (2004) FEBS Lett. 564, 312]. Because fluorinated and hydrogenated chains do not mix well, supramicellar concentrations of these surfactants can coexist with intact lipid ... More
Multicolor single-molecule FRET to explore protein folding and binding.
Authors:Gambin Y, Deniz AA,
Journal:Mol Biosyst
PubMed ID:20601974
'Proper protein function in cells, tissues and organisms depends critically on correct protein folding or interaction with partners. Over the last decade, single-molecule FRET (smFRET) has emerged as a powerful tool to probe complex distributions, dynamics, pathways and landscapes in protein folding and binding reactions, leveraging its ability to avoid ... More