Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • ATPIF1 Antibodies

          Invitrogen

          ATPIF1 Monoclonal Antibody (5E2D7)

          2 Published Figures
          12 References
          View all (12) ATPIF1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite ATPIF1 Monoclonal Antibody (5E2D7)

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (24)
          • Published Figures (2)
          ATPIF1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          ATPIF1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 26

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          ATPIF1 Antibody (A-21355) in ICC/IF

          CAKI cells were plated on coverslips overnight. The next day cells were fixed and permeabilized using the Image-iT® Fixation/Permeabilization Kit (Product # R37602) according to protocol. Following suppression of background binding using Image-iT® FX Signal enhancer (Product # R37107), cells were incubated with 3 µg/mL anti-ATP synthase subunit IF1 (Product # A-21355) for labeling of mitochondria for 30 min at room temperature and washed three times with dPBS, followed by Alexa Fluor® 594 goat anti-rabbit IgG antibody ReadyProbes® reagent ... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          ATPIF1 Antibody in Immunocytochemistry (ICC/IF)
          ATPIF1 Antibody in Immunocytochemistry (ICC/IF)
          ATPIF1 Antibody in Immunocytochemistry (ICC/IF)
          ATPIF1 Antibody in Immunocytochemistry (ICC/IF)
          ATPIF1 Antibody in Immunocytochemistry (ICC/IF)
          ATPIF1 Antibody in Immunocytochemistry (ICC/IF)
          ATPIF1 Antibody in Immunocytochemistry (ICC/IF)
          ATPIF1 Antibody in Immunocytochemistry (ICC/IF)
          ATPIF1 Antibody in Immunocytochemistry (ICC/IF)
          ATPIF1 Antibody in Immunocytochemistry (ICC/IF)
          ATPIF1 Antibody in Immunocytochemistry (ICC/IF)
          ATPIF1 Antibody in Immunocytochemistry (ICC/IF)
          ATPIF1 Antibody in Immunocytochemistry (ICC/IF)
          ATPIF1 Antibody in Immunocytochemistry (ICC/IF)
          ATPIF1 Antibody in Immunocytochemistry (ICC/IF)
          ATPIF1 Antibody in Immunocytochemistry (ICC/IF)
          ATPIF1 Antibody in Immunocytochemistry (ICC/IF)
          ATPIF1 Antibody in Immunocytochemistry (ICC/IF)
          ATPIF1 Antibody in Immunocytochemistry (ICC/IF)
          ATPIF1 Antibody in Immunocytochemistry (ICC/IF)
          ATPIF1 Antibody in Immunocytochemistry (ICC/IF)
          ATPIF1 Antibody in Immunocytochemistry (ICC/IF)
          ATPIF1 Antibody in Immunocytochemistry (ICC/IF)
          ATPIF1 Antibody in Immunocytochemistry (ICC/IF)
          ATPIF1 Antibody in Immunocytochemistry (ICC/IF)
          ATPIF1 Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          A-21355

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1 µg/mL
          View 2 publications 2 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          -
          View 1 publication 1 publication

          Immunohistochemistry (Frozen) (IHC (F))

          -
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          1-5 µg/mL
          View 6 publications 6 publications

          Proximity Ligation Assay (PLA) (PLA)

          -
          View 1 publication 1 publication

          Miscellaneous PubMed (Misc)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Bovine, Human, Mouse, Rat

          Published species

          Bovine, Dog, Human, Mouse, Non-human primate, Rat

          Host/Isotype

          Mouse / IgG1, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          5E2D7

          Immunogen

          Recombinant IF1.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          purified

          Storage buffer

          PBS, pH 7.4

          Contains

          0.02% sodium azide

          Storage conditions

          4°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2535841

          Product Specific Information

          The antibody was produced in vitro using hybridomas grown in serum-free medium, then purified by biochemical fractionation, and is near homogeneity as judged by SDS-PAGE.

          This antibody recognizes bands at ~10 and ~18 kDa on western analysis.

          Target Information

          Complex V, also called F1F0 ATPase or ATP synthase is extremely conserved through evolution and can be found in plants, fungi, bacteria, and animals. The ATP synthase enzyme is a transmembrane protein responsible for driving the reversible reaction from ADP + phosphate to ATP, in oxidative phosphorylation, and as a proton pumping ATPase. This reaction is accomplished by a flux of protons across the membrane as a result of electron transfer. The enzyme was thought to be localized exclusively to mitochondria; however, it has recently been identified on the plasma membrane of several cell types including hepatocytes where it functions as the HDL receptor, on endothelial cells where it may act as the angiostatin receptor, and on the surface of cancer cells. The ATP synthase protein has two main sections; the F1 ATP-ase (soluble) and the F0 ATP-ase (membrane embedded). The F1 section consists of the alpha, beta, gamma, delta, and epsilon subunits; while the F0 consists of a, b, and c subunits. The enzyme in mammals is composed of 17 subunits, five of which make up the easily detached F1. The remainder subunits are components of two stalk domains and the proton pumping F0 part of the machinery. Two of the subunits of the F0 part are encoded on mitochondrial DNA while the other subunits are nuclearly encoded. Mutations in the mitochondrial-encoded subunits of ATP synthase (Complex V) cause OXPHOS disease.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: ATP synthase F1 subunit epsilon; ATP synthase inhibitor protein; ATPase inhibitor (rat mitochondrial IF1 protein); ATPase inhibitor protein; ATPase inhibitory factor 1; ATPIF1; ATPIP; IF(1); IF1; inhibitor for F1F0-ATPase; inhibitor of F(1)F(o)-ATPase; unnamed protein product

          View more View less

          Gene Aliases: ATPI; ATPIF1; ATPIP; IF(1); If1; IF1PA; IP

          View more View less

          UniProt ID: (Human) Q9UII2, (Bovine) P01096, (Mouse) O35143, (Rat) Q03344

          View more View less

          Entrez Gene ID: (Human) 93974, (Bovine) 327699, (Mouse) 11983, (Rat) 25392

          View more View less

          Function(s)
          enzyme inhibitor activity protein binding calmodulin binding enzyme binding ATPase inhibitor activity identical protein binding angiostatin binding ATPase binding mitochondrial proton-transporting ATP synthase complex binding structural molecule activity protein homodimerization activity ligase
          Process(es)
          angiogenesis negative regulation of endothelial cell proliferation response to ischemia generation of precursor metabolites and energy heme biosynthetic process negative regulation of cardiac muscle cell apoptotic process erythrocyte differentiation positive regulation of protein catabolic process negative regulation of hydrolase activity mitochondrial depolarization regulation of protein localization to mitochondrion positive regulation of type 2 mitophagy negative regulation of mitochondrial ATP synthesis coupled proton transport protein homotetramerization reactive oxygen species metabolic process positive regulation of mitochondrial outer membrane permeabilization involved in apoptotic signaling pathway regulation of ATP metabolic process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-frpqk:80/100.66.72.150:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch