Pluripotent Stem Cell Immunocytochemistry Kit (OCT4, SSEA4) - FAQs
Pluripotent Stem Cell Immunocytochemistry Kit (OCT4, SSEA4) - FAQs
View additional product information for Pluripotent Stem Cell Immunocytochemistry Kit (OCT4, SSEA4) - FAQs (A25526)
3 product FAQs found
The Pluripotent Stem Cell Immunocytochemistry Kit provides the NucBlue Fixed Cell Stain. If I don't have the correct filter set for this stain, can I use another nucleic acid stain?
If you use Alexa Fluor 488/Alexa Fluor 555 dye-conjugated secondary antibodies and you have a Cy5/Alexa Fluor 647 filter set, you may use a far-red nucleic acid stain, such as NucRed Dead 647 ReadyProbes Reagent (Cat. No. R37113) or SYTOX Deep Red Nucleic Acid Stain (Cat. No. S11380). Do not use these far-red nucleic acid stains if using an Alexa Fluor 594 dye-conjugated secondary antibody as there is too much spectral overlap of Alexa Fluor 594 into the Cy5/Alexa Fluor 647 range.
Can I use the Pluripotent Stem Cell Immunocytochemistry Kit for flow cytometry analysis?
We do not recommend using the Pluripotent Stem Cell Immunocytochemistry Kit for flow cytometry analysis. The kit and protocol were specifically developed for imaging analysis (microscopy/high-content screening (HCS)).
Do you offer the individual components of the Pluripotent Stem Cell Immunocytochemistry Kit as standalone products?
Only the NucBlue Fixed Cell ReadyProbes Reagent may be purchased separately as Cat. No. R37606. We do offer the secondary antibodies separately, but not in the same proprietary concentration as provided in the kit. We also offer formaldehyde, DPBS, BSA, and saponin as separate catalog numbers, but not in the volumes or as formulated in the kit. The primary antibodies are not available as separate stock items; all non-stock items in the kit may be available as a custom order. Please contact your Account Representative.