PowerUp™ SYBR™ Green Master Mix for qPCR
Applied Biosystems™

PowerUp™ SYBR™ Green Master Mix for qPCR

Pre-formulated, optimized, universal 2X master mix for real-time PCR workflows.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A2574350 mL
A257411 mL
A257425 mL
A2577810 x 5 mL
A257805 x 1 mL
A2591810 x 1 mL
A257762 x 5 mL
A257775 x 5 mL
A257792 x 1 mL
Catalog number A25743
Price (USD)
2,565.00
Special offer
Online exclusive
Ends: 11-Dec-2026
4,750.00
Save 2,185.00
Each
Quantity:
50 mL
Price (USD)
2,565.00
Special offer
Online exclusive
Ends: 11-Dec-2026
4,750.00
Save 2,185.00
Each

PowerUp™ SYBR™ Green Master Mix is a pre-formulated, optimized, universal 2X master mix for real-time PCR workflows. Coupled with user-supplied primer sets and template, PowerUp™ SYBR™ Green Master Mix is designed to amplify targets for accurate gene expression analysis.

PowerUp™ SYBR™ Green Master Mix features include:

  • A dual hot-start mechanism for excellent specificity
  • Highly reproducible CTs over a broad dynamic range
  • Inclusion of UDG to help prevent carryover contamination
  • Stability of pre-assembled reactions for up to 72 hours
  • Compatibility with most real-time qPCR instruments

Exceptional specificity with dual hot-start mechanism

Specificity is paramount in obtaining high-quality data in SYBR™ Green reactions and can be enhanced through control of the Taq DNA polymerase at lower temperatures before stringent primer binding. PowerUp™ SYBR™ Green Master Mix uses the Dual-Lock™ Taq DNA polymerase enzyme that combines two unique hot-start mechanisms to help control its activity and to prevent undesirable early activity of the polymerase at low temperatures. In an experiment evaluating 24 different primer sets, PowerUp™ SYBR™ Green Master Mix generated single melt curves 100% of the time, consistent with highly specific amplification.

Tight reproducibility in CTs over a wide dynamic range

PowerUp™ SYBR™ Green Master Mix demonstrates excellent reproducibility over a wide dynamic range for a variety of targets tested (see figure). In addition, this master mix provides efficient amplification over 6 logs of sample input. Unlike competitor mixes that can exhibit inhibition at high cDNA inputs and low correlation coefficients over a dynamic range experiment, PowerUp™ SYBR™ Green Master Mix efficiency is maintained over the full dynamic range to help deliver maximal confidence in data quality (see figure).

UDG included for carryover contamination control

The inclusion of uracil-DNA glycosylase (UDG) and dUTP in the PowerUp™ SYBR™ Green Master Mix enables the degradation of any previously amplified PCR products, helping prevent contamination of subsequent qPCR reactions and possible false positives.

Stability of pre-assembled reactions

The tight control of the Dual-Lock™ Taq DNA polymerase in PowerUp™ SYBR™ Green Master Mix enables reactions to be assembled up to 72 hours prior to cycling without impacting data (see figure). This consistency allows users to have confidence and flexibility in the use of PowerUP™ SYBR™ Green Master Mix across numerous workflow scenarios¹.

Broad instrument compatibility

PowerUp™ SYBR™ Green Master Mix can be used in either standard or fast cycling mode and is compatible with all Applied Biosystems™ real-time PCR instruments². It is also compatible with the Bio-Rad IQ™5 and CFX96™ / CFX384™ systems, Roche LightCycler™ LC480, and Stratagene™ MX3005P™ systems. For optimal results, recommended primer concentrations are 300–800 nM.

¹Pre-PCR stability is influenced not only by the master mix, but also by the target being analyzed. For maximum confidence, researchers should confirm stability profiles of their specific targets.

² For optimal performance, standard cycling using the Applied Biosystems™ 7900 system is recommended.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
For Use With (Equipment)7500 Fast System, 7500 System, 7900HT System, QuantStudio 12k Flex, QuantStudio 3, QuantStudio 5, QuantStudio 6 Flex, QuantStudio 6 Pro, QuantStudio 7 Pro, QuantStudio 7 Flex, StepOne, StepOnePlus, ViiA 7 System, Roche LightCycler PRO System, Bio-Rad CFX Opus 96 Real-Time PCR System, Applied Biosystems AmpQuest 96
FormatTube
IncludesdUTP, UNG/UDG
Inhibitor Tolerance (Simple)Medium
Multiplex CapabilitySingle-plex
No. of Reactions5000 Reactions
Passive Reference DyeROX (Pre-mixed)
PolymeraseDual-Lock Taq DNA Polymerase
Product LinePowerUP, SYBR
Product TypeReal Time PCR SYBR Master Mix
Quantity50 mL
Sample TypeDNA
Shipping ConditionWet Ice
Sufficient For5000 Reactions of 20 μL
Target SpecificitycDNA or gDNA only
Thermostability72 hr., Assembled Reaction
Volume50 mL
Concentration2X
Detection MethodSYBR
For Use With (Application)Gene Expression
PCR MethodqPCR
Reaction SpeedFast or Standard
Unit SizeEach
Contents & Storage
The 2X mix contains SYBR Green dye, Dual-Lock Taq DNA Polymerase, dNTPs with dUTP/dTTP blend, heat-labile UDG, ROX passive reference dye, and optimized buffer components. Contains 1 x 50 mL tube, sufficient for 5000 20-μL reactions (10 μL Master Mix per reaction).

Store at 2–8°C in the dark.

Frequently asked questions (FAQs)

When using the PowerUp SYBR Green Master Mix (Cat. No. A25741, A25742, A25743, A25776, A25777, A25778, A25779, A25780, A25919), I get high Ct values. What can I do?

It could be due to the following reasons:

  • Insufficient cDNA template is present. Use 10 to 100 ng of cDNA template per 50 µL reaction.
  • Quality of cDNA template is poor. Quantify the amount of cDNA template and test the cDNA template for the presence of PCR inhibitors.
  • Sample degradation. Prepare fresh cDNA, then repeat the experiment.
  • Reduced number of PCR cycles in the thermal cycling protocol. Increase the number of PCR cycles to the default setting of 40.
  • Primer-dimer formation and residual polymerase activity. Prepare the reaction mixes and reaction plate on ice. To ensure optimal results, run the reaction plate as soon as possible after completing the reaction setup.


Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Applications Support Center.

When using the PowerUp SYBR Green Master Mix (Cat. No. A25741, A25742, A25743, A25776, A25777, A25778, A25779, A25780, A25919), the Multicomponent signal for ROX is not linear. What could cause this?

This could be due to the following reasons:

  • Pure dye component’s spectra are incorrect. Rerun pure dye spectra.
  • Incorrect dye components have been chosen. Choose correct dyes for data analysis.


Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Applications Support Center.

Which reverse transcription kit can I use with the PowerUp SYBR Green Master Mix (Cat. No. A25741, A25742, A25743, A25776, A25777, A25778, A25779, A25780, A25919)?

We recommend using the High-Capacity RNA-to-cDNA Kit (Cat. No. 4387406, 4388950).

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Applications Support Center.

What primer concentrations can I use with the PowerUp SYBR Green Master Mix (Cat. No. A25741, A25742, A25743, A25776, A25777, A25778, A25779, A25780, A25919)?

The recommended primer concentration range is 300-800 nM. You will need to optimize the primer concentration by running different concentration combinations of the forward and reverse primer. See page 28 of the PowerUp SYBR Green Master Mix User Guide.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Applications Support Center.

Does the PowerUp SYBR Green Master Mix (Cat. No. A25741, A25742, A25743, A25776, A25777, A25778, A25779, A25780, A25919) contain uracil-DNA glycolase?

This master mix has a heat-labile UDG. It is completely and irreversibly inactivated after 10 minute incubation at 50°C.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Applications Support Center.