QuantStudio™ 3D Digital PCR Master Mix v2
Applied Biosystems™

QuantStudio™ 3D Digital PCR Master Mix v2

QuantStudio 3D Digital PCR MasterMix v2 is designed to deliver accurate, sensitive, and precise digital PCR data.Read more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A263581.5 mL
A263595 mL
Catalog number A26358
Price (MXN)
-
Quantity:
1.5 mL
QuantStudio 3D Digital PCR MasterMix v2 is designed to deliver accurate, sensitive, and precise digital PCR data.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)QuantStudio™
Product LineQuantStudio
Product TypeDigital PCR Master Mix
Quantity1.5 mL
Shipping ConditionDry Ice
Detection MethodPrimer-probe
For Use With (Application)Digital PCR (dPCR)
FormLiquid
PCR MethodDigital PCR
Unit SizeEach
Contents & Storage
Store at -5°C to -30°C until first use and then store (at 2°C to 8°C.)

Frequently asked questions (FAQs)

How do I replace the fuses in the QuantStudio 3D Digital PCR System?

Please follow the procedures described in "Chapter A: Maintenance" of the QuantStudio 3D Digital PCR System User Guide to replace the fuses. Make sure to only replace with those listed and certified fuses of the same type and rating as those currently in the QuantStudio 3D instrument.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.

How does sample preparation for digital PCR differ from real-time PCR?

The answer is that sample prep for digital PCR is no different from real-time PCR when using the QuantStudio 3D Digital PCR System. If you have samples in your freezer that you've run on real-time PCR, then you're ready to run them with digital PCR.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.

How do I determine how much sample to use when setting up the digital PCR reaction for the QuantStudio 3D Digital PCR System?

Please see the section on Sample Dilution in Chapter 3 of the QuantStudio 3D Digital PCR System User Guide (http://tools.thermofisher.com/content/sfs/manuals/MAN0007720.pdf) for more details on how to determine the target copy number per genome.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.

Where are my chip image data saved on the QuantStudio 3D instrument?

You can configure the QuantStudio 3D instrument to upload the imaging data to a USB drive, a network file server, or the cloud data storage service. See Appendix B, “Networking”, in the QuantStudio 3D Digital PCR System User Guide for more information on setting up network destinations. Also, the QuantStudio 3D instrument retains a copy of the analyzed data from the imaged QuantStudio 3D Digital PCR 20K Chip that you can access from the Run History screen. Note: The results will remain in the QuantStudio 3D instrument cache for approximately 300 readings. After 300 chips, the instrument removes the oldest data file in the cache to store each new reading.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.

Is there any way to access the raw fluorescence data from the QuantStudio 3D Chips in AnalysisSuite software v3.0?

Yes, there is an option to export the Chip data as .xml on the Export tab in AnalysisSuite software v3.0.