Avidin, NeutrAvidin™, PE conjugate
Invitrogen™

Avidin, NeutrAvidin™, PE conjugate

Our R-phycoerythrin (R-PE) conjugate of NeutrAvidin® biotin-binding protein - a form of avidin that has been processed to remove carbohydrate자세히 알아보기
Have Questions?
카탈로그 번호수량
A26601 mL
카탈로그 번호 A2660
제품 가격(KRW)
590,000
온라인 행사
Ends: 30-Sep-2026
693,000
할인액 103,000 (15%)
Each
수량:
1 mL
제품 가격(KRW)
590,000
온라인 행사
Ends: 30-Sep-2026
693,000
할인액 103,000 (15%)
Each
Our R-phycoerythrin (R-PE) conjugate of NeutrAvidin® biotin-binding protein - a form of avidin that has been processed to remove carbohydrate and to lower its isoelectric point - can substantially decrease background due to nonspecific binding. The method used to deglycosylate the avidin retains its specific binding. This bright, fluorescent reagent is ideal for detecting biotinylated targets in flow cytometry applications.
For Research Use Only. Not for use in diagnostic procedures.
사양
농도1 mg/mL
제품 유형Avidin
수량1 mL
배송 조건Wet Ice
콘주게이트R-phycoerythrin
제품라인NeutrAvidin
Unit SizeEach
구성 및 보관
Store in refrigerator (2–8°C) and protect from light.

인용 및 참조 문헌 (7)

인용 및 참조 문헌
Abstract
Detection of kappa opioid receptors on mouse thymocyte phenotypic subpopulations as assessed by flow cytometry.
Authors:Ignatowski TA, Bidlack JM
Journal:J Pharmacol Exp Ther
PubMed ID:9435191
'Recent studies have shown kappa opioid receptor labeling on the R1EGO thymoma cell line by indirect immunofluorescence and flow cytometric analysis. The present study used a fluorescein-labeled arylacetamide (FITC-AA), a kappa opioid ligand, in conjunction with biotin-conjugated anti-fluorescein IgG and extravidin-R-phycoerythrin (PE), along with double-labeling with antibodies against specific immune ... More
A complementary role for the tetraspanins CD37 and Tssc6 in cellular immunity.
Authors:Gartlan KH, Belz GT, Tarrant JM, Minigo G, Katsara M, Sheng KC, Sofi M, van Spriel AB, Apostolopoulos V, Plebanski M, Robb L, Wright MD,
Journal:J Immunol
PubMed ID:20709950
'The cooperative nature of tetraspanin-tetraspanin interactions in membrane organization suggests functional overlap is likely to be important in tetraspanin biology. Previous functional studies of the tetraspanins CD37 and Tssc6 in the immune system found that both CD37 and Tssc6 regulate T cell proliferative responses in vitro. CD37(-/-) mice also displayed ... More
Improved immunocytochemical detection of biotinylated probes with Neutralite avidin.
Authors:van Roy N, Mangelschots K, Speleman F
Journal:Trends Genet
PubMed ID:8488564
Isolating and engineering human antibodies using yeast surface display.
Authors:Chao G, Lau WL, Hackel BJ, Sazinsky SL, Lippow SM, Wittrup KD
Journal:Nat Protoc
PubMed ID:17406305
This protocol describes the process of isolating and engineering antibodies or proteins for increased affinity and stability using yeast surface display. Single-chain antibody fragments (scFvs) are first isolated from an existing nonimmune human library displayed on the yeast surface using magnetic-activated cell sorting selection followed by selection using flow cytometry. ... More
High-throughput single-molecule spectroscopy in free solution.
Authors:Ma Y, Shortreed MR, Yeung ES
Journal:Anal Chem
PubMed ID:11028623
A high-speed high-throughput single-molecule imaging technique for identifying molecules in free solution based on differences in their fluorescence emission spectra is presented. Unlike previous reports, the entire spectrum, rather than selected wavelengths through optical filters, is recorded. Furthermore, the millisecond data acquisition time means that the molecules do not need ... More